Skip to Content
Merck
All Photos(1)

Key Documents

I2778

Sigma-Aldrich

IL-1α human

≥98% (SDS-PAGE and HPLC), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

IL-1α, hIL-1α., Interleukin-1α human, Lymphocyte activating factor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Interleukin-1α human, IL-1α, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE and HPLC)

form

lyophilized powder

quality

endotoxin tested

mol wt

~17 kDa

packaging

pkg of 2 and 10 μg

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL1A(3552)

General description

Interleukin-1α (IL-1α) is a cytokine that belongs to the interleukin-1 (IL-1) family. It is located intracellularly in hematopoietic and non-hematopoietic cells. IL-1α gene is located on the human chromosome at 2q14.1.

Application

Interleukin-1α (IL-1α) has been used:
  • to study its effects on human colorectal adenocarcinoma cells
  • to stimulate primary human fetal retinal pigment epithelium (hfRPE) cells
  • to pre-stimulate human THP-1 monocytes

Biochem/physiol Actions

Interleukin-1α (IL-1α) plays a crucial role in inflammation during cell necrosis. It also regulates the recruitment of interleukin-1β (IL-1β), neutrophils, chemokines, and other cytokines at the site of injury. IL-1α exhibits a dual function of binding to its cell surface receptor as well as chromatin.

Physical form

Lyophilized from 0.2 μm filtered 10 mM sodium phosphate , pH 8.0, plus 50 mm NaCl and 50 μg bovine serum albumin (BSA) per μg cytokine.

Analysis Note

The proliferative activity of human IL-1α is tested in culture using the murine cell line D10S.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nelson C Di Paolo et al.
Nature immunology, 17(8), 906-913 (2016-07-21)
Inflammation occurs after disruption of tissue homeostasis by cell stress, injury or infection and ultimately involves the recruitment and retention of cells of hematopoietic origin, which arrive at the affected sites to resolve damage and initiate repair. Interleukin 1α (IL-1α)
Niklas Palmqvist et al.
Journal of inflammation (London, England), 13, 20-20 (2016-06-25)
Atopic dermatitis (AD) is a chronic inflammatory skin disease characterized by highly pruritic eczematous lesions that are commonly treated with topical corticosteroids and calcineurin inhibitors. Side-effects and safety concerns associated with these agents restrict their use, and new, safe treatment
Ivo C J H Post et al.
Xenotransplantation, 20(1), 18-26 (2013-01-26)
Endothelial damage is a critical step in the development of (xeno) transplantation-related and cardiovascular pathology. In humans, the amount of circulating endothelial cells (CEC) correlates to disease intensity and functions as a valuable damage marker. While (xeno) transplantation and cardiovascular
Association of interleukin1 beta, interleukin1 receptor antagonist, interleukin 6 and angiotensin II receptor type I gene polymorphisms with end stage renal disease: A meta-analysis
Divya M, et al.
Gene Reports, 100856-100856 (2020)
Peleg Rider et al.
Seminars in immunology, 25(6), 430-438 (2013-11-05)
Although the IL-1α molecule has long been recognized, information about its distinct role in various diseases is limited, since most clinical studies have focused on the role of IL-1β. Despite triggering the same IL-1 receptor as does IL-1β, there is

Articles

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service