Skip to Content
Merck
All Photos(3)

Key Documents

HPA032148

Sigma-Aldrich

Anti-ANO1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DOG1, Anti-FLJ10261, Anti-ORAOV2, Anti-TAOS2, Anti-TMEM16A, Anti-anoctamin 1, calcium activated chloride channel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

RVNEKYSTLPAEDRSVHIINICAIEDIGYLPSEGTLLNSLSVDPDAECKYGLYFRDGRRKVDYILVYHHKRPSGNRTLVRRVQHSDTPSGARSVKQDHP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANO1(55107)

General description

Anoctamin 1 (ANO1), a membrane protein, is a member of a protein family with eight transmembrane helices and N- and C-termini. It has two conserved domains, a meiotic segregation-interfering domain, and multiple glycosylation and phosphorylation sites. ANO1 gene consists of 26 exons and is mapped to human chromosome 11q13.

Immunogen

anoctamin 1, calcium activated chloride channel recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-ANO1 antibody produced in rabbit has been used: in immunofluorescence(1:100) via confocal microscopy and western blot (1:1000).

Biochem/physiol Actions

Anoctamin 1 (ANO1) protein serves as a calcium-activated chloride channel (CaCC). It is essential for the normal development of the trachea. ANO1 plays a key role in cell motility, invasion, and adhesion of head and neck squamous cell carcinoma (HNSCC) cells. It might be a promising target for anticancer therapy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78510

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hari P Dhakal et al.
The American journal of surgical pathology, 43(8), 1123-1128 (2019-05-17)
Evaluation of testicular biopsies from azoospermic men requires recognition of phases of germ cell maturation as organized architecturally within the seminiferous tubule, as well as distinguishing the inability to generate mature spermatozoa (germ cell aplasia or maturation arrest) from normal
Zongtao Liu et al.
International journal of cancer, 144(9), 2215-2226 (2018-09-23)
Most common ovarian cancers are epithelial carcinoma in which the etiology for carcinogenesis remains elusive. ANO1/TMEM16A, a member of Ca2+ -activated Cl- channels (CaCCs), has been demonstrated to promote epithelium-originated cancers and whether it plays a role in the pathogenesis
C Ayoub et al.
British journal of cancer, 103(5), 715-726 (2010-07-29)
Head and neck squamous cell carcinoma (HNSCC) is associated with poor survival. To identify prognostic and diagnostic markers and therapeutic targets, we studied ANO1, a recently identified calcium-activated chloride channel (CaCC). High-resolution genomic and transcriptomic microarray analysis and functional studies
Fangzhen Wang et al.
Experimental and therapeutic medicine, 17(1), 298-306 (2019-01-18)
Network-based systems biology has become an important method for analysis of high-throughput gene expression data and gene function mining. The aim of the present study was to implement a weighted gene co-expression network analysis to screen genes that were significantly
Fiona M Bryant et al.
The Plant cell, 31(6), 1276-1288 (2019-04-10)
Seed dormancy governs the timing of germination, one of the most important developmental transitions in a plant's life cycle. The DELAY OF GERMINATION1 (DOG1) gene is a key regulator of seed dormancy and a major quantitative trait locus in Arabidopsis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service