Skip to Content
Merck
All Photos(2)

Key Documents

HPA018507

Sigma-Aldrich

Anti-TMED1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Interleukin-1 receptor-like 1 ligand, Anti-Putative T1/ST2 receptor- binding protein, Anti-Transmembrane emp24 domain-containing protein 1 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PPIQDGEFTFLLPAGRKQCFYQSAPANASLETEYQVIGGAGLDVDFTLESPQGVLLVSESRKADG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TMED1(11018)

General description

TEMED1 (transmembrane emp24 domain-containing protein-1) belongs to TMED/p24 family of proteins. TEMED1 is widely expressed. The protein is present in endoplasmic reticulum, cis-Golgi network and endoplasmic reticulum-Golgi intermediate compartment. The protein exists as a homodimer.

Immunogen

Transmembrane emp24 domain-containing protein 1 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-TMED1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Transmembrane emp24 domain-containing protein-1 (TEMED1) is important for innate immune signaling. TMED1 interacts with IL (Interleukin)-1 receptor family member ST2L and optimizes IL-33-induced IL-8 and IL-6 production.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71706

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Gayle et al.
The Journal of biological chemistry, 271(10), 5784-5789 (1996-03-08)
T1/ST2 is a receptor-like molecule homologous to the type I interleukin-1 receptor. Despite this sequence similarity, we have been unable to demonstrate binding of T1/ST2 to any of the three interleukin-1 species. In searching for a ligand for T1/ST2, we
Nicole Jenne et al.
The Journal of biological chemistry, 277(48), 46504-46511 (2002-09-19)
The p24 proteins belong to a highly conserved family of membrane proteins that cycle in the early secretory pathway. They bind to the coat proteins of COPI and COPII vesicles, and are proposed to be involved in vesicle biogenesis, cargo
Dympna J Connolly et al.
The Journal of biological chemistry, 288(8), 5616-5623 (2013-01-16)
The proinflammatory danger signal IL-33, which is released from damaged or dying cells, achieves its effects via the IL-1R family member ST2L. The detection of IL-33 by ST2L initiates downstream signaling pathways that result in the activation of MAPKs and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service