Skip to Content
Merck
All Photos(2)

Key Documents

HPA017976

Sigma-Aldrich

Anti-PCDH18 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Protocadherin-18 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

IHKGDITLVPTINGTLPIRSHHRSSPSSSPTLERGQMGSRQSHNSHQSLNSLVTISSNHVPENFSLELTHATPAVEQVSQLLSMLHQGQYQPRPSFRGNKYSRSYR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCDH18(54510)

General description

Protocadherin 18 (PCDH18) is part of the cadherin superfamily. It is located at the synapses in the central nervous system and contains six ectodomain repeats with cadherin-like features. The gene encoding PCDH18 is localized on human chromosome 4.

Immunogen

Protocadherin-18 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-PCDH18 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Protocadherin 18 (PCDH18) is associated with synaptic functions and takes part in maintaining cell to cell connections in the brain. It acts as an activation marker of CD8+ memory T cells. A deletion in the gene has been associated with developmental delay in the brain.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72391

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinical and Molecular Delineation of a Novel De Novo 4q28.3-31.21 Interstitial Deletion in a Patient with Developmental Delay.
John Hoon Rim et al.
Yonsei medical journal, 56(6), 1742-1744 (2015-10-09)
Sarah L Kerns et al.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology, 107(3), 372-376 (2013-05-31)
Rectal bleeding can occur following radiotherapy for prostate cancer and negatively impacts quality of life for cancer survivors. Treatment and clinical factors do not fully predict rectal bleeding, and genetic factors may be important. A genome-wide association study (GWAS) was
Jurate Kasnauskiene et al.
European journal of medical genetics, 55(4), 274-277 (2012-03-28)
We report a boy with severe developmental delay, seizures, microcephaly, hypoplastic corpus callosum, internal hydrocephalus and dysmorphic features (narrow forehead, round face, deep-set eyes, blue sclerae, large and prominent ears, nose with anteverted nares, thin upper lip, small and wide-spaced
Edwin J Vazquez-Cintron et al.
PloS one, 7(5), e36101-e36101 (2012-05-09)
CD8(+) tumor infiltrating T cells (TIL) lack effector-phase functions due to defective proximal TCR-mediated signaling previously shown to result from inactivation of p56(lck) kinase. We identify a novel interacting partner for p56(lck) in nonlytic TIL, Protocadherin-18 ('pcdh18'), and show that
T Wolverton et al.
Genomics, 76(1-3), 66-72 (2001-09-11)
Protocadherins are members of a nonclassic subfamily of calcium-dependent cell-cell adhesion molecules in the cadherin superfamily. Although the extracellular domains have several common structural features, there is no extensive homology between the cytoplasmic domains of protocadherin subfamily members. We have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service