Skip to Content
Merck
All Photos(5)

Key Documents

HPA017876

Sigma-Aldrich

Anti-KTN1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CG-1 antigen, Anti-Kinectin, Anti-Kinesin receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:1000- 1:2500

immunogen sequence

ELKRLEAMLKERESDLSSKTQLLQDVQDENKLFKSQIEQLKQQNYQQASSFPPHEELLKVISEREKEISGLWNELDSLKDAVEHQRKKNNERQQQVEAVELEAKE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KTN1(3895)

General description

Kinectin 1 (KTN1) is a 156kDa receptor to which kinesin (a motor protein) binds. It contains a central domain and a RhoA binding domain in its carboxyl terminus. KTN1 is present in the endoplasmic reticulum membrane and the gene encoding it is localized on human chromosome 14q22.1.

Immunogen

Kinectin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Kinectin 1 (KTN1) enhances the ATPase activity of kinesin and is also a crucial mediator of RhoG (a GTPase) cellular activity. It binds to the GTP-bound form of RhoG. KTN1 functions as a kinesin anchor protein and is important for the vesicle movement along microtubules. It also binds and anchors the elongation factor-1 complex onto the endoplasmic reticulum and thus plays a key role in regulating protein synthesis in cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70517

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinectin is a key effector of RhoG microtubule-dependent cellular activity.
Vignal E, et al.
Molecular and Cellular Biology, 21(23), 8022-8034 (2001)
Kinectin is a key effector of RhoG microtubule-dependent cellular activity.
E Vignal
Molecular and Cellular Biology, 21(23), 8022-8034 (2001)
Assignment of the human kinectin gene (KTN1), encoding a kinesin-binding protein, to chromosome 14 band q22.1 by in situ hybridization.
P N Rao et al.
Cytogenetics and cell genetics, 79(3-4), 196-197 (1997-01-01)
K Salassidis et al.
Cancer research, 60(11), 2786-2789 (2000-06-13)
Evaluation of 20 cases of radiation-induced childhood papillary thyroid carcinoma using fluorescence in situ hybridization demonstrated the presence of clonal translocations affecting the RET locus. Semiquantitative reverse transcription-PCR indicated overexpression of the RET tyrosine kinase (TK) domain in four cases.
Lee-Lee Ong et al.
The Journal of biological chemistry, 281(44), 33621-33634 (2006-09-05)
Kinectin is an integral membrane protein with many isoforms primarily found on the endoplasmic reticulum. It has been found to bind kinesin, Rho GTPase, and translation elongation factor-1delta. None of the existing models for the quaternary organization of the elongation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service