Skip to Content
Merck
All Photos(3)

Key Documents

HPA012891

Sigma-Aldrich

Anti-DSC1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DG2/DG3, Anti-Desmocollin-1 precursor, Anti-Desmosomal glycoprotein 2/3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

SCTGTLVVHLDDYNDHAPQIDKEVTICQNNEDFAVLKPVDPDGPENGPPFQFFLDNSASKNWNIEEKDGKTAILRQRQNLDYNYYSVPIQIKDRHGLVATHMLTVRVCDCSTPSECRMKDKSTR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DSC1(1823)

General description

Desmocollin-1 precursor (DSC1) is a cadherin-related glycoprotein present in desmosomes. It belongs to the desmosomal cadherin family. The gene encoding this protein is localized on chromosome 18.

Immunogen

Desmocollin-1 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-DSC1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Desmocollin-1 precursor (DSC1) plays an important role in cell-cell adhesion. It maintains tissue integrity in epithelial cells and cardiac muscles.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71939

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I A King et al.
Genomics, 18(2), 185-194 (1993-11-01)
Desmosomes are adhesive epithelial junctions that contain two distinct classes of cadherin-related glycoproteins (desmogleins and desmocollins), both of which occur as several different isoforms whose expression is related to epithelial differentiation. We have now isolated cDNA clones encoding a human
Linda Paavilainen et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 58(3), 237-246 (2009-11-11)
Pathology archives harbor large amounts of formalin-fixed, paraffin-embedded tissue samples, used mainly in clinical diagnostics but also for research purposes. Introduction of heat-induced antigen retrieval has enabled the use of tissue samples for extensive immunohistochemical analysis, despite the fact that
K Khan et al.
British journal of cancer, 95(10), 1367-1370 (2006-11-08)
The desmocollins are members of the desmosomal cadherin family of cell-cell adhesion molecules. They are essential constituents of desmosomes, intercellular junctions that play a critical role in the maintenance of tissue integrity in epithelia and cardiac muscle. In humans, three
J A McGovern et al.
The British journal of dermatology, 176(1), 145-158 (2016-07-22)
Epidermal homeostasis is maintained through the balance between keratinocyte proliferation, differentiation and desquamation; however, human skin equivalent (HSE) models are known to differentiate excessively. In native tissue, proteases such as kallikrein-related peptidase (KLK) 5 and KLK7 cleave the extracellular components

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service