Skip to Content
Merck
All Photos(4)

Key Documents

HPA010512

Sigma-Aldrich

Anti-CLPS antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Colipase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:2500-1:5000

immunogen sequence

NLENGELCMNSAQCKSNCCQHSSALGLARCTSMASENSECSVKTLYGIYYKCPCERGLTCEGDKTIVGSITNTNFGICHDAGR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLPS(1208)

General description

CLPS (colipase) is a low molecular weight pancreatic secretory protein. This protein is composed of 112 amino acids, which contains a 17-amino acid leader peptide. This gene is localized to human chromosome 6, which is composed of three exons.

Immunogen

Colipase Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CLPS (colipase) functions as a cofactor for pancreatic triglyceride lipase for the efficient hydrolysis of dietary lipid. Its expression is increased by secretin and dietary fats. Arg92Cys polymorphism in this gene is linked with increased susceptibility to type II diabetes. This polymorphism reduces functionality against long-chain triglycerides, which might be a possible cause of type II diabetes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71985

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H van Tilbeurgh et al.
Biochimica et biophysica acta, 1441(2-3), 173-184 (1999-11-26)
Colipase is a small protein cofactor needed by pancreatic lipase for the efficient dietary lipid hydrolysis. It binds to the C-terminal, non-catalytic domain of lipase, thereby stabilising an active conformation and considerably increasing the overall hydrophobic binding site. Structural studies
M E Lowe et al.
Biochemistry, 29(3), 823-828 (1990-01-23)
Pancreatic lipase hydrolyzes dietary triglycerides to monoglycerides and fatty acids. In the presence of bile salts, the activity of pancreatic lipase is markedly decreased. The activity can be restored by the addition of colipase, a low molecular weight protein secreted
H F Sims et al.
Biochemistry, 31(31), 7120-7125 (1992-08-11)
The digestion of dietary triglycerides occurs in the duodenum through the action of triglyceride lipase, a pancreatic exocrine protein. The activity of pancreatic lipase is inhibited by the bile salts normally found in the gut lumen. Another pancreatic exocrine protein
Xunjun Xiao et al.
Journal of lipid research, 54(2), 514-521 (2012-12-04)
Colipase is essential for efficient fat digestion. An arginine-to-cysteine polymorphism at position 92 of colipase (Arg92Cys) associates with an increased risk for developing type-2 diabetes through an undefined mechanism. To test our hypothesis that the extra cysteine increases colipase misfolding
Sheryl D'Silva et al.
Journal of lipid research, 48(11), 2478-2484 (2007-08-24)
Type 2 diabetes mellitus is a multifactorial and polygenic disorder with increasing prevalence. Recently, a polymorphism in the gene encoding procolipase, a cysteine for arginine substitution at position 92, was associated with type 2 diabetes in two human populations. Because

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service