Skip to Content
Merck
All Photos(2)

Key Documents

HPA008773

Sigma-Aldrich

Anti-CA12 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CA-XII antibody produced in rabbit, Anti-Carbonate dehydratase XII antibody produced in rabbit, Anti-Carbonic anhydrase 12 precursor antibody produced in rabbit, Anti-Carbonic anhydrase XII antibody produced in rabbit, Anti-Tumor antigen HOM-RCC-3.1.3 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

TASNKSEGLAVLAVLIEMGSFNPSYDKIFSHLQHVKYKGQEAFVPGFNIEELLPERTAEYYRYRGSLTTPPCNPTVLWTVFRNPVQISQEQLLALETALYCTHMDDPSPREMINNFRQVQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CA12(771)

General description

CA12 (carbonic anhydrase XII) gene is localized to human chromosome 15q22. This gene is predominantly expressed in adult pancreas, colon, kidney and prostate. It belongs to α CA gene family, and is composed of a leader sequence, an α CA domain, a transmembrane region and a short cytoplasmic domain.

Immunogen

Carbonic anhydrase 12 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-CA12 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CA12 (carbonic anhydrase XII) catalyzes the hydration of CO2 generated by cells into bicarbonate and protons. Expression of this enzyme in oral squamous cell carcinoma (OSCC) is related to tumor progression and poor prognosis. The expression levels of this enzyme are also linked to the histological grade of cervical cancer, and absence of its expression correlates with poorly differentiated phenotype. The presence of this enzyme marginally links with better disease-free survival of cervical cancer patients.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71649

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeroen F Vermeulen et al.
BMC cancer, 12, 240-240 (2012-06-15)
Mammographic population screening in The Netherlands has increased the number of breast cancer patients with small and non-palpable breast tumors. Nevertheless, mammography is not ultimately sensitive and specific for distinct subtypes. Molecular imaging with targeted tracers might increase specificity and
Mark C Lloyd et al.
Cancer research, 76(11), 3136-3144 (2016-03-25)
Spatial heterogeneity in tumors is generally thought to result from branching clonal evolution driven by random mutations that accumulate during tumor development. However, this concept rests on the implicit assumption that cancer cells never evolve to a fitness maximum because
Krissie Lenting et al.
Acta neuropathologica communications, 7(1), 185-185 (2019-11-22)
Many biology-based precision drugs are available that neutralize aberrant molecular pathways in cancer. Molecular heterogeneity and the lack of reliable companion diagnostic biomarkers for many drugs makes targeted treatment of cancer inaccurate for many individuals. Identifying actionable hyperactive biological pathways
Karen A Power et al.
Arthritis and rheumatism, 63(12), 3876-3886 (2011-12-01)
Back pain is a major cause of disability, affecting millions of people worldwide. One cause of axial back pain is degeneration of the nucleus pulposus (NP) of the intervertebral disc. This study was undertaken to investigate associations of NP cells
Chong Woo Yoo et al.
Radiation oncology (London, England), 5, 101-101 (2010-11-03)
To investigate whether expression of carbonic anhydrase XII (CA12) is associated with histologic grade of the tumors and radiotherapy outcomes of the patients with invasive cervical cancer. CA12 expression was examined by immunohistochemical stains in cervical cancer tissues from 183

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service