Skip to Content
Merck
All Photos(4)

Key Documents

HPA005895

Sigma-Aldrich

Anti-MUC4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ASGP antibody produced in rabbit, Anti-Ascites sialoglycoprotein antibody produced in rabbit, Anti-Mucin-4 precursor antibody produced in rabbit, Anti-Pancreatic adenocarcinoma mucin antibody produced in rabbit, Anti-Testis mucin antibody produced in rabbit, Anti-Tracheobronchial mucin antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

IQYTSNAEDANFTLRDSCTDLELFENGTLLWTPKSLEPFTLEILARSAKIGLASALQPRTVVCHCNAESQCLYNQTSRVGNSSLEVAGCKCDGGTFGRYCEGSEDACEEPCFPSVHCVPGKGCEACPPNLTGD

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUC4(4585)

Immunogen

Mucin-4 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Mucin 4 (MUC 4) is a mucin protein encoded by the MUC4 gene in humans. It is referred as ASGP, MUC-4 and HSA276359. It is a transmembrane glycoprotein that is expressed in pancreatic ductal adenocarcinoma (PDAC). It has a role in pancreatic tumor progression and metastasis. It plays an important role in epithelial cell proliferation and differentiation. Its overexpression is associated with invasive tumor proliferation and several cancers. It possesses tumor-promoter functions. It is a high molecular weight membrane-bound transmembrane glycoprotein that is frequently detected in invasive and metastatic cancer. It is found to play a critical role in regulating EMT by modulating β-catenin. This gene may function as a prognostic biomarker and could be a potential therapeutic target for lung adenocarcinoma metastasis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70005

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liuwei Gao et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(2), 1335-1341 (2013-09-17)
The mucin MUC4 is a high molecular weight membrane-bound transmembrane glycoprotein that is frequently detected in invasive and metastatic cancer. The overexpression of MUC4 is associated with increased risks for several types of cancer. However, the functional role of MUC4
Zili Zhang et al.
PloS one, 8(10), e77723-e77723 (2013-11-10)
Mucin MUC4, which is encoded by the MUC4 gene, plays an important role in epithelial cell proliferation and differentiation. Aberrant MUC4 overexpression is associated with invasive tumor proliferation and poor outcome in epithelial cancers. Collectively, the existing evidence suggests that
Daniel Ansari et al.
Scandinavian journal of gastroenterology, 48(10), 1183-1187 (2013-09-21)
OBJECTIVE. Mucin 4 (MUC4) is a transmembrane glycoprotein that is expressed in pancreatic ductal adenocarcinoma (PDAC), but not in normal pancreatic tissue. MUC4 has a proposed role in pancreatic tumor progression and metastasis. The purpose of this pilot study was
Rajani Rai et al.
Data in brief, 48, 109243-109243 (2023-06-29)
The objective was to find an association between abnormal glycosylation, represented by Tn and STn antigens on mucin (MUC) proteins, in primary tumor specimens with lymph node metastasis or recurrence of cervical cancer patients. Prospectively collected specimens were obtained from

Protocols

Subcellular protein location often correlates with function; nucleus proteins implicated in gene regulation, mitochondria proteins in energy production.

Subcellular protein location often correlates with function; nucleus proteins implicated in gene regulation, mitochondria proteins in energy production.

Subcellular protein location often correlates with function; nucleus proteins implicated in gene regulation, mitochondria proteins in energy production.

Subcellular protein location often correlates with function; nucleus proteins implicated in gene regulation, mitochondria proteins in energy production.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service