Skip to Content
Merck
All Photos(5)

Key Documents

HPA001939

Sigma-Aldrich

Anti-MMP2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-72 kDa gelatinase antibody produced in rabbit, Anti-72 kDa type IV collagenase precursor antibody produced in rabbit, Anti-Gelatinase A antibody produced in rabbit, Anti-MMP-2 antibody produced in rabbit, Anti-Matrix metalloproteinase-2 antibody produced in rabbit, Anti-TBE-1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50-1:200
western blot: 0.04-0.4 μg/mL

immunogen sequence

YGASPDIDLGTGPTPTLGPVTPEICKQDIVFDGIAQIRGEIFFFKDRFIWRTVTPRDKPMGPLLVATFWPELPEKIDAVYEAPQEEKAVFFAGNEYWIYSASTLERGYPKPLTSLGLPPDVQRVDAAFNWS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP2(4313)

General description

MMP2 gene is located on the human chromosome at 16q12.2.
Matrix metalloproteinase 2 (MMP2) belong to the homologous matrix metalloproteinase (MMP) family. It consists of a homologous cysteine-switch and zinc binding regions. It is synthesized by proliferating skin fibroblasts, specific bronchial epithelial cells, and SV40-transformed lung fibroblasts.

Immunogen

72 kDa type IV collagenase precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-MMP2 antibody produced in rabbit has been used in western blotting and immunohistochemistry.
Anti-MMP2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

MMP2 (matrix metalloproteinase 2) is involved in cleaving triple helical type I and type IV collagen as well as degradation of denatured collagens. Mutation of MMP2 enzymatic activity in the serum or fibroblasts leads to the multicentric osteolysis and arthritis syndrome.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83038

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Guang Shen et al.
Oncology letters, 13(3), 1119-1124 (2017-04-30)
Esophageal carcinoma is one of the most common malignancies in China. Previous studies reported that matrix metalloproteinases (MMPs) have important roles in the progression and invasion of numerous types of solid tumors. Among the MMPs, MMP-2 has been closely associated
Matrix metalloproteinase-2 and-9 expression in the epiligament of the medial collateral and anterior cruciate ligament in human knees: a comparative study
Georgiev G P, et al.
Cureus, 10(11), 17-26 (2018)
Sándor Farkas et al.
The Journal of biological chemistry, 299(9), 104998-104998 (2023-07-03)
Chlorotoxin (CTX), a scorpion venom-derived 36-residue miniprotein, binds to and is taken up selectively by glioblastoma cells. Previous studies provided controversial results concerning target protein(s) of CTX. These included CLC3 chloride channel, matrix metalloproteinase 2 (MMP-2), regulators of MMP-2, annexin
Georgi P Georgiev et al.
Cureus, 10(11), e3550-e3550 (2019-01-17)
Aim Ninety percent of knee ligament injuries involve the medial collateral ligament (MCL) and the anterior cruciate ligament (ACL) of the knee joint. Matrix metalloproteinases (MMPs) are a large group of calcium- and zinc-dependent endopeptidases responsible for cleaving and rebuilding
MMP-2,-3 and TIMP-2,-3 polymorphisms in colorectal cancer in a Chinese Han population: A case-control study
Wang N, et al.
Gene, 730, 144320-144320 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service