Skip to Content
Merck
All Photos(1)

Key Documents

H9408

Sigma-Aldrich

β-Hydroxybutyrate Dehydrogenase from Pseudomonas lemoignei

lyophilized powder, ≥200 units/mg protein

Synonym(s):

(R)-3-Hydroxybutanoate:NAD+ oxidoreductase, 3-HBDH

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (Pseudomonas lemoignei)

Quality Level

form

lyophilized powder

specific activity

≥200 units/mg protein

purified by

chromatography

foreign activity

lactic dehydrogenase ≤0.05%
malic dehydrogenase ≤0.1%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

D-β-hydroxybutyrate dehydrogenase (BDH) is a membrane bound lipid-requiring enzyme.

Application

β-Hydroxybutyrate Dehydrogenase from Pseudomonas lemoignei has been used in coupled assay to confirm the formation of acetoacetate.
Suitable for the determination of acetoacetate and D(-)-3-hydroxybutyrate by the method of Williamson, D. H., and Mellanby, J., Methods of Enzymatic Analysis, Bergmeyer, H., ed., 2nd edition, 4, 1836 (1974).

Biochem/physiol Actions

In Paracoccus denitrificans, 3-hydroxybutyrate dehydrogenase plays a key role in the degradation of intracellular polyhydroxybutyrate and polyhydroxyvalerate.
In mammalian systems, β-hydroxybutyrate dehydrogenase is localized on the inner mitochondrial membrane and requires phosphatidyl choline for activity. In contrast, the enzyme from Pseudomonas is a soluble cytosolic enzyme that does not require a phospholipid allosteric activator. The enzyme is required for the utilization of ketone bodies as a source of metabolic energy. It catalyzes the oxidation of 3-hydroxybutyrate to acetoacetate, the first step in the conversion of ketone bodies to citric acid, which is then further metabolized via the tricarboxylic acid cycle (Krebs cycle).

Unit Definition

One unit will oxidize 1.0 μmole of D-β-hydroxybutyrate to acetoacetate per min at pH 7.8 at 37 °C.

Physical form

lyophilized powder containing sucrose, β-NAD and Tris buffer salts

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D-beta-HYDROXYBUTYRATE DEHYDROGENASE: A MOLECULAR BIOLOGY APPROACH TO THE STUDY OF A LIPID-REQUIRING ENZYME
Structure and Function of Membrane Proteins, 283-300 (1983)
3-Hydroxybutyrate oligomer hydrolase and 3-hydroxybutyrate dehydrogenase participate in intracellular polyhydroxybutyrate and polyhydroxyvalerate degradation in Paracoccus denitrificans
Lu J, et al.
Applied and Environmental Microbiology, 80(3), 986-993 (2014)
The specific molecular architecture of plant 3-hydroxy-3-methylglutaryl-CoA lyase
Hemmerlin A, et al.
The Journal of biological chemistry, 80(3) (2019)
J Nie et al.
Scandinavian journal of medicine & science in sports, 21(5), 625-629 (2010-05-13)
This study examined the response of serum biomarkers of cardiac and skeletal muscle damage at rest and after a routine workout of 21 km run in 12 male adolescent (16.2±0.6 years) long-distance runners. Biomarkers of cardiac [troponins (cTnT, cTnI), creatine
Michel A Sciotti et al.
Chimia, 64(11), 793-798 (2011-01-05)
Gamma hydroxybutyric acid (GHB) is a regulated therapeutic drug, which naturally occurs in mammalian brain tissues as an intermediate of the GABA (gamma aminobutyric acid) neurotransmitter metabolism. The increasing misuse of GHB as a narcotic or abusing drug in recent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service