Skip to Content
Merck
All Photos(1)

Key Documents

H7660

Sigma-Aldrich

Heregulin-β1 (EGF Domain) human

≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder

Synonym(s):

HRG-β1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

<0.50 ng/mL

mol wt

predicted mol wt 8 kDa

packaging

pkg of 10 and 50 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

color

white

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... NRG1(3084)

Biochem/physiol Actions

Growth factor ligand for ErbB3 and ErbB4 receptor tyrosine kinases. The binding of HRG results in receptor dimerization and receptor trans-autophosphorylation. The phosphorylated receptors recruit cellular signaling proteins, initiating signaling pathways.

Analysis Note

The biological activity is measured by its ability to stimulate the proliferation of human MCF-7 cells grown under serum-free conditions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Tan et al.
The Journal of biological chemistry, 282(33), 24343-24351 (2007-06-15)
Neuregulin 1 (NRG1) is essential for the development and function of multiple organ systems, and its dysregulation has been linked to diseases such as cancer and schizophrenia. Recently, altered expression of a novel isoform (type IV) in the brain has
A M Addington et al.
Molecular psychiatry, 12(2), 195-205 (2006-10-13)
Childhood-onset schizophrenia (COS), defined as onset of psychosis by the age of 12, is a rare and malignant form of the illness, which may have more salient genetic influence. Since the initial report of association between neuregulin 1 (NRG1) and
Neural grafts containing exosomes derived from Schwann cell-like cells promote peripheral nerve regeneration in rats.
Hu, et al.
Burns & trauma, 11, tkad013-tkad013 (2023)
Shuji Wakatsuki et al.
Genes to cells : devoted to molecular & cellular mechanisms, 19(1), 66-77 (2013-11-22)
After peripheral nerve injury, Schwann cells gain a migratory phenotype and remodel their extracellular matrix to provide a supportive environment for axonal regeneration. The soluble neuregulin-1 isoform, that is, glial growth factor (GGF), is expressed in regenerating axons of injured
Paul J Harrison et al.
Biological psychiatry, 60(2), 132-140 (2006-01-31)
Neuregulin 1 (NRG1) is a leading schizophrenia susceptibility gene. The NRG1 locus on chromosome 8p shows linkage to the disorder, and genetic association has been found between schizophrenia and various non-coding polymorphisms and haplotypes, especially at the 5' end of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service