Skip to Content
Merck
All Photos(1)

Key Documents

H3917

Sigma-Aldrich

Heparinase I and III Blend from Flavobacterium heparinum

lyophilized powder, stabilized with ∼ 25% (w/w) bovine serum albumin, ≥200 unit/mg protein (enzyme + BSA)

Synonym(s):

Heparinase I and Heparinase III blend

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (Flavobacterium heparinum)

Quality Level

conjugate

conjugate (Glucosaminoglycan)

form

lyophilized powder

specific activity

≥200 units/mg protein

concentration

≥200 unit/mg protein (enzyme + BSA)

shipped in

dry ice

storage temp.

−20°C

General description

Heparinase is an inducible, non-extracellular heparin-degrading enzyme. Three types of heparinises are produced by Flavobacterium heparinum and contains specific sequences of heparin.

Application

Heparinase I and III Blend from Flavobacterium heparinum has been used in:
  • the digestion of heparan sulfate from ovine vitreous
  • human embryonic kidney cells
  • glycosaminoglycans from arterial tissues
  • P0 retinae digestion

Biochem/physiol Actions

Heparinase I and III plays vital role in various biological processes: modulate cell-growth factor interactions, cell-lipoprotein interactions, neovascularization. It cleaves highly sulphated polysaccharide chains in presence of 2-O-sulfated α-L-idopyranosyluronic acid and β-D-glucopyranosyluronic acid residues of polysaccharides.
Heparin-degrading lyase that recognizes heparin sulfate proteoglycan as its primary substrate.

Packaging

Sold on the basis of Heparinase I units

Unit Definition

One unit will form 0.1 micromole of unsaturated uronic acid per hour at 7.5 at 25 degrees C using Heparin, Sodium as substrate for heparinase I.

One unit will form 0.1 micromole of unsaturated uronic acid per hour at 7.5 at 25 degrees C using bovine kidney Heparan, Sulfate as substrate for heparinase III.

One unit will form 0.1 μmole of unsaturated uronic acid per hr at pH 7.5 at 25 °C. One International Unit (I.U.) is equivalent to approx. 600 Sigma units. Package sizes are sold in Sigma units.

Other Notes

Enzyme Commission Numbers: 4.2.2.7 Hep I and 4.2.2.8 Hep III

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glycosaminoglycans contribute to extracellular matrix fiber recruitment and arterial wall mechanics
Mattson JM, et al.
Biomechanics and Modeling in Mechanobiology, 16(1), 213-225 (2017)
R Sasisekharan et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(4), 1524-1528 (1994-02-15)
Neovascularization is associated with the regulation of tissue development, wound healing, and tumor metastasis. A number of studies have focused on the role of heparin-like molecules in neovascularization; however, little is known about the role of heparin-degrading enzymes in neovascularization.
IL-2 inducible kinase ITK is critical for HIV-1 infection of Jurkat T-cells
Hain A, et al.
Scientific reports, 8(1), 3217-3217 (2018)
Retinal proteoglycans act as cellular receptors for basement membrane assembly to control astrocyte migration and angiogenesis
Tao C and Zhang X
Cell Reports, 17(7), 1832-1844 (2016)
Diffusion regulation in the vitreous humor
Kasdorf B, et al.
Biophysical Journal, 109(10), 2171-2181 (2015)

Articles

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service