Skip to Content
Merck
All Photos(3)

Key Documents

H3413

Sigma-Aldrich

Anti-Histone Deacetylase 10 (HDAC10) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HD10

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~72 kDa

species reactivity

mouse, rat, human

technique(s)

indirect immunofluorescence: 10-20 μg/mL using human HeLa cells
microarray: suitable
western blot (chemiluminescent): 0.5-1.0 μg/mL using whole extracts of mouse NIH3T3 and rat NRK cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Histone deacetylase 10 (HDAC10) is highly expressed in liver, kidney, pancreas and spleen. HDAC10 is similar to HDAC6, both containing a unique putative second catalytic domain not found in other HDACs. HDAC10 is localized to both the nucleus and cytoplasm. Histone deacetylases (HDACs) are competing enzymes, belonging to histone deacetylase family. HDAC10 is a novel member of the class II family of HDACs with a bipartite structure consisting of Hda1p-related catalytic domain at the N-terminal and a leucine-rich domain at the C-terminal. It is present, both in the nucleas and cytoplasm.

Immunogen

synthetic peptide corresponding to amino acid residues 2-16 of human HDAC10 with C-terminal added cysteine, conjugated to KLH.

Application

Anti-Histone Deacetylase 10 (HDAC10) antibody produced in rabbit has been used in western blot analysis and immunofluorescence.

Biochem/physiol Actions

Histone deacetylase 10 (HDAC10) can deacetylate histones, repress transcription and interact with HDAC3. HDAC10 can stimulate lung cancer proliferation via AKT phosphorylation. It is involved in homologous recombination.
The leucine-rich domain on the C-terminal of HDAC8 is responsible for cytoplasmic enrichment. Trichostatin A (TSA), a specific antitumor deacetylase inhibitor is sensitive to the enzymatic activity of HDAC10. HDAC10 attaches to a promoter and represses transcription in the nucleas.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salma Darwish et al.
International journal of molecular sciences, 23(14) (2022-07-28)
In addition to involvement in epigenetic gene regulation, histone deacetylases (HDACs) regulate multiple cellular processes through mediating the activity of non-histone protein substrates. The knockdown of HDAC8 isozyme is associated with the inhibition of cell proliferation and apoptosis enhancement in
Polyamine Homeostasis in Snyder-Robinson Syndrome
Murray-ST, et al.
Medical sciences (Basel, Switzerland), 6(4), 112-112 (2018)
Dual role of HDAC10 in lysosomal exocytosis and DNA repair promotes neuroblastoma chemoresistance
Ridinger J, et al.
Scientific reports, 8(1), 10039-10039 (2018)
Tracy Murray-Stewart et al.
Medical sciences (Basel, Switzerland), 6(4) (2018-12-14)
Loss-of-function mutations of the spermine synthase gene (SMS) result in Snyder-Robinson Syndrome (SRS), a recessive X-linked syndrome characterized by intellectual disability, osteoporosis, hypotonia, speech abnormalities, kyphoscoliosis, and seizures. As SMS catalyzes the biosynthesis of the polyamine spermine from its precursor
Annunziata Gloghini et al.
British journal of haematology, 147(4), 515-525 (2009-09-25)
Unselective histone deacetylase (HDAC) inhibitors are a promising novel therapy for lymphoid malignancies. However, these treatments remain empiric as the pattern of HDAC enzymes in different types of cancer, including lymphoid malignancies, remains unknown. We examined the expression of class

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service