Skip to Content
Merck
All Photos(1)

Key Documents

H0913

Sigma-Aldrich

Anti-acetyl-Histone H3 (Ac-Lys9) antibody, Mouse monoclonal

clone AH3-120, purified from hybridoma cell culture

Synonym(s):

Anti-H3K9ac

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AH3-120, monoclonal

form

buffered aqueous solution

mol wt

antigen ~17 kDa

species reactivity

human, bovine, Caenorhabditis elegans, frog, Drosophila, chicken, rat, mouse

packaging

antibody small pack of 25 μL

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: 1-2 μg/mL using whole cell extract of mouse fibroblasts 3T3 cell line treated with sodium butyrate

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

acetylation (Lys9)

General description

Monoclonal Anti-Acetyl-Histone H3 (Ac-Lys9) recognizes human histone H3 when acetylated on Lys9. Staining of the histone H3-Ac-Lys9 band in immunoblotting is specifically inhibited with the acetylated histone H3 immunizing peptide but not with the nonacetylated one.

Immunogen

synthetic, acetylated histone H3 peptide (amino acids 7-20, Ac-Lys9) corresponding to the N-terminus of human histone H3. The sequence is identical in many species including mouse, rat, bovine, chicken, frog, Drosophila, and C. elegans, and is highly conserved (single amino acid substitution) in Tetrahymena histone H3.

Application

Monoclonal Anti-acetyl-Histone H3 (Ac-Lys9) antibody may be used in various applications including ELISA, immunoblotting (approx. 17 kDa), and immunocytochemistry.

Biochem/physiol Actions

Acetyl-Histone H3 hav Acetylation of lysine residues within these N-terminal domains of histones by histone acetyl-transferase (HATs), including Gcn5p, PCAF, p300/CBP and TAFII250 is associated with transcriptional activation. This modification results in remodeling of the nucleosome structure into an open conformation more accessible to transcription complexes. Conversely, histone deacetylation by histone deacetylase (HDACs) is associated with transcription repression reversing the chromatin remodeling process. In most species, histone H3 is primarily acetylated at lysine 9, 14, 18, and 23. Acetylation at lysine 9 appears to have a dominant role in histone deposition and chromatin assembly in some organisms. Acetylation of specific lysines in histone H3 is also associated with processes apart from transcription. During DNA replication, new histones are rapidly synthesized and assembled into replicated DNA. Histones H3 and H4 are brought to replicating chromatin in a pre-acetylated state that turns into a de-acetylated state after replication is completed and the newly assembled chromatin matures.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel Arabidopsis acetyltransferase interacts with the geminivirus movement protein NSP
McGarry RC, et al.
Plant Cell, 15, 1605-1618 (2003)
Regulation of immune responses by histone deacetylase inhibitor
Licciardi PV and Karagiannis TC
ISRN hematology, 2012 (2012)
Priyanka Jain et al.
Scientific reports, 9(1), 8508-8508 (2019-06-13)
Glycosylphosphatidylinositol (GPI)-anchored proteins are important for virulence of many pathogenic organisms including the human fungal pathogen, Candida albicans. GPI biosynthesis is initiated by a multi-subunit enzyme, GPI-N-acetylglucosaminyltransferase (GPI-GnT). We showed previously that two GPI-GnT subunits, encoded by CaGPI2 and CaGPI19
Histone H3 variants and modifications on transcribed genes
Workman JL and Abmayr SM
Proceedings of the National Academy of Sciences of the USA, 101, 1429-1430 (2004)
Parisa Nadri et al.
PloS one, 17(7), e0267598-e0267598 (2022-07-22)
SCNT embryos suffer from poor developmental competence (both in vitro and in vivo) due to various defects such as oxidative stress, incomplete epigenetic reprogramming, and flaws in telomere rejuvenation. It is very promising to ameliorate all these defects in SCNT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service