Skip to Content
Merck
All Photos(2)

Key Documents

G9753

Sigma-Aldrich

α-D-Glucosamine 1-phosphate

Synonym(s):

2-Amino-2-deoxy-α-D-glucopyranosyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14NO8P
CAS Number:
Molecular Weight:
259.15
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

natural (inorganic)

Assay

≥97% (TLC)

form

powder

impurities

<8.5% water (Karl Fischer)

color

white

solubility

water: 100 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

NC1C(O)C(O)C(CO)OC1OP(O)(O)=O

InChI

1S/C6H14NO8P/c7-3-5(10)4(9)2(1-8)14-6(3)15-16(11,12)13/h2-6,8-10H,1,7H2,(H2,11,12,13)

InChI key

YMJBYRVFGYXULK-UHFFFAOYSA-N

Application


  • Cellodextrin phosphorylase from Ruminiclostridium thermocellum: X-ray crystal structure and substrate specificity analysis. This study presents the enzymatic synthesis and analysis of alpha-ᴅ-Glucosamine 1-phosphate based polysaccharides using cellodextrin phosphorylase, showcasing potential for novel biomaterial development. Field et al., 2017

  • Glucose-1-phosphate uridylyltransferase from Erwinia amylovora: Activity, structure and substrate specificity. This paper explores the biochemical pathway involving alpha-ᴅ-Glucosamine 1-phosphate in the context of bacterial metabolism, providing insights into microbial biochemistry and potential targets for antibacterial therapy. Field et al., 2017

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Hatzack et al.
Journal of chromatography. B, Biomedical sciences and applications, 736(1-2), 221-229 (2000-02-17)
A simple and inexpensive high-performance thin-layer chromatography (HPTLC) method for the analysis of inositol mono- to hexakisphosphates on cellulose precoated plates is described. Plates were developed in 1-propanol-25% ammonia solution-water (5:4:1) and substance quantities as low as 100-200 pmol were
K Takayama et al.
The Journal of biological chemistry, 258(12), 7379-7385 (1983-06-25)
We have determined the complete structure of a glycolipid (designated lipid X) previously found to accumulate in certain Escherichia coli mutants defective in phosphatidylglycerol synthesis (Nishijima, M., and Raetz, C.R.H. (1979) J. Biol. Chem. 254, 7837-7844). Based on fast atom
S Ambrosio et al.
Journal of biochemical and biophysical methods, 25(4), 237-244 (1992-12-01)
Galactosamine is quickly metabolized to galactosamine 1-phosphate in rats treated with this compound. An HPLC method to quantify hexosamine phosphates in biological samples is described, modified from the o-phthaldialdehyde amino acid analysis procedure. o-Phthaldialdehyde derivatives of hexosamines and hexosamine-phosphates can
E V Vorob'eva et al.
Bioorganicheskaia khimiia, 32(5), 538-545 (2006-10-18)
The hydrolysis of defatted cells of the marine bacterium Chryseobacterium scophtalmum CIP 104199T with 10% acetic acid (3 h, 100 degrees C) led to an unusual lipid A (LA) (yield 0.6%), obtained for the first time. Using chemical analysis, FAB
Seema C Namboori et al.
Journal of bacteriology, 190(8), 2987-2996 (2008-02-12)
Archaea and eukaryotes share a dolichol phosphate-dependent system for protein N-glycosylation. In both domains, the acetamido sugar N-acetylglucosamine (GlcNAc) forms part of the core oligosaccharide. However, the archaeal Methanococcales produce GlcNAc using the bacterial biosynthetic pathway. Key enzymes in this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service