Skip to Content
Merck
All Photos(3)

Key Documents

G5544

Sigma-Aldrich

Anti-GABAA Receptor (α6 subunit) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-γ-Aminobutyric Acid Type A Receptor (α6 subunit), Anti-GABRA6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot: 1:200 using rat brain membranes

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GABRA6(2559)
mouse ... Gabra6(14399)
rat ... Gabra6(29708)

General description

GABAA and GABAB receptors differ with regard to their ionic characteristics and pharmacological properties. The GABAA receptor is an ionotropic receptor that forms the GABA gated chloride channel and consists of several heterogeneous subunits with membrane recognition sites for benzodiazapenes.

Immunogen

peptide corresponding to amino acids 20-37 of rat GABA(A) α6. This sequence is identical in mouse and shares 17 of 19 residues in human.

Application

Anti-GABAA Receptor (α 6 subunit) antibody produced in rabbit is suitable for the immunofluorescent detection of GABAA receptor α 6 in cerebella of mice. It is suitable for immunoblotting at a working dilution of 1:200 using rat brain membranes.

Biochem/physiol Actions

The inhibitory neurotransmitter GABA signals through two distinct types of pre- and postsynaptic receptors, GABAA and GABAB. Both GABA receptors can regulate depression of synaptic transmission and be involved in the inhibition controlling neuronal excitability. Mutations in the gene may be responsible for the pathogenesis of childhood absence epilepsy (CAE). Polymorphism in this gene is also related to attenuation in hormonal and blood pressure responses under certain psychological stress.

Target description

Anti-GABA(A) Receptor (α6 subunit) specifically recognizes the GABA(A) receptor α6 subunit protein and may be used for the detection of the GABA(A) receptor α6 subunit protein. The inhibitory neurotransmitter GABA (α-aminobutyricacid

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanne Falck et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 40(14), 2943-2959 (2020-03-04)
Piccolo, a presynaptic active zone protein, is best known for its role in the regulated assembly and function of vertebrate synapses. Genetic studies suggest a further link to several psychiatric disorders as well as Pontocerebellar Hypoplasia type 3 (PCH3). We
Ciria C Hernandez et al.
The Journal of physiology, 589(Pt 23), 5857-5878 (2011-09-21)
A GABA(A) receptor α6 subunit mutation, R46W, was identified as a susceptibility gene that may contribute to the pathogenesis of childhood absence epilepsy (CAE), but the molecular basis for alteration of GABA(A) receptor function is unclear. The R46W mutation is
Erick J Rodríguez-Palma et al.
Pain, 164(5), 948-966 (2022-08-25)
The loss of GABAergic inhibition is a mechanism that underlies neuropathic pain. Therefore, rescuing the GABAergic inhibitory tone through the activation of GABA A receptors is a strategy to reduce neuropathic pain. This study was designed to elucidate the function
M Uhart et al.
Molecular psychiatry, 9(11), 998-1006 (2004-06-16)
The glucocorticoid component of the stress response has been the subject of intense scientific scrutiny because of the wide ranging pathological consequences resulting from excess glucocorticoid exposure, including mood and anxiety disorders, and cognitive impairment. Exposure to stress activates the
Fabian M P Kaiser et al.
The Journal of experimental medicine, 219(9) (2022-08-11)
The genetic causes of primary antibody deficiencies and autism spectrum disorder (ASD) are largely unknown. Here, we report a patient with hypogammaglobulinemia and ASD who carries biallelic mutations in the transcription factor PAX5. A patient-specific Pax5 mutant mouse revealed an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service