Skip to Content
Merck
All Photos(1)

Key Documents

G5500

Sigma-Aldrich

L-(−)-Glucose

≥99%

Synonym(s):

Aldehydo-L-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724626
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99%

form

powder

technique(s)

gas chromatography (GC): suitable

color

white

mp

153-156 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

[H]C(=O)[C@@H](O)[C@H](O)[C@@H](O)[C@@H](O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4+,5+,6+/m1/s1

InChI key

GZCGUPFRVQAUEE-VANKVMQKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-(−)-Glucose is an enantiomer of the more common D-glucose. L-Glucose cannot be used by cells as an energy source because it cannot be phosphorylated by hexokinase.

Application

L-(-)-Glucose has been used in phospho-buffered saline (PBS) solution to induce metabolic responses in Saccharomyces cerevisiae. It has also been used to study the substrate competition pattern of IICB glucose transporter.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A metabolic interpretation for the response of cellular autofluorescence to chemical perturbations assessed using spectral phasor analysis
Maltas J, et al.
Royal Society of Chemistry Advances, 8(72), 41526-41535 (2018)
Cheung Pang Wong et al.
Cells, 8(7) (2019-07-18)
Zika virus (ZIKV) has emerged as an important human pathogen that can cause congenital defects in the fetus and neurological conditions in adults. The interferon (IFN) system has proven crucial in restricting ZIKV replication and pathogenesis. The canonical IFN response
Measuring substrate binding and affinity of purified membrane transport proteins using the scintillation proximity assay
Harder D and Fotiadis D
Nature Protocols, 7(9), 1569-1569 (2012)
Amitai D Mandelbaum et al.
Diabetologia, 62(9), 1653-1666 (2019-06-13)
Adult beta cells in the pancreas are the sole source of insulin in the body. Beta cell loss or increased demand for insulin impose metabolic challenges because adult beta cells are generally quiescent and infrequently re-enter the cell division cycle.
Dan-Dan Huang et al.
Shock (Augusta, Ga.), 51(6), 745-756 (2018-06-05)
Sirtuin1 (Sirt1) and Sirtuin3 (Sirt3) are known to participate in regulating mitochondrial function. However, whether Total Salvianolic Acid Injection (TSI) protects against myocardial ischemia/reperfusion (I/R) injury through regulating Sirt1, Sirt3, and mitochondrial respiratory chain complexes is unclear. The aim of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service