Skip to Content
Merck
All Photos(1)

Key Documents

F3013

Sigma-Aldrich

Ferredoxin from Spinacia oleracea (spinach)

lyophilized powder, Ferredoxin ≥15 wt. %

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

spinach (Spinacia oleracea)

Quality Level

form

lyophilized powder

composition

Ferredoxin, ≥15 wt. %

solubility

water: 2 mg/mL, clear, faintly brown

application(s)

cell analysis

storage temp.

−20°C

Gene Information

spinach ... SOVF(137530)

Looking for similar products? Visit Product Comparison Guide

General description

Ferredoxin I (Fd I) from Spinacia oleracea comprises 97 amino acid and is an iron sulfur cluster (2Fe-2S). It is synthesized in the chloroplast. The structure of Fd I has conserved active site.

Application

Ferredoxin from Spinacia oleracea (spinach) has been used:
  • as a substrate for spinach ferredoxin reductase,
  • for reconstitution with chlorophyll-A for scanning electron microscope (SEM) dispersive X-ray analyzer (EDX) integrated analysis,
  • as a reductant in adenylylsulfate (APS) reductase activity of P. aeruginosa

Biochem/physiol Actions

Ferredoxin is important in the electron transfer cascade required for this nitrogen assimilation. It has been used in studies of assimilation of inorganic nitrogen from nitrite or nitrate in the absence of ammonium.
Ferredoxins are iron-containing proteins that serve as electron-transfer catalysts in photosynthesis and nitrogen fixation.

Packaging

Package sizes are based on ferredoxin content.

Physical form

Contains Tris buffer

Preparation Note

Partially purified by the method of Tagawa and Arnon, Nature, 195, 537 (1962).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keita Miyake et al.
The FEBS journal, 287(18), 4016-4031 (2020-01-30)
Bilin pigments play important roles for both light perception and harvesting in cyanobacteria by binding to cyanobacteriochromes (CBCRs) and phycobilisomes (PBS), respectively. Among various cyanobacteria, Acaryochloris marina MBIC 11017 (A. marina 11017) exceptionally uses chlorophyll d as the main photosynthetic pigment absorbing
Harnessing a P450 fatty acid decarboxylase from Macrococcus caseolyticus for microbial biosynthesis of odd chain terminal alkenes
Lee JW, et al.
Metabolic Engineering Communications, 7, e00076-e00076 (2018)
Structure of the Mutant E92K of [2Fe-2S] Ferredoxin I from Spinacia oleracea at 1.7 AA Resolution
Binda C, et al
Acta Crystallographica Section D, Biological Crystallography, 54(6), 1353-1358 (1998)
Growth-promoting effect on iron-sulfur proteins on axenic cultures of Entamoeba dispar
Khalifa SAM, et al.
Parasite (Paris, France), 13(1), 51-58 (2006)
Extraction of ferredoxin from spinach leaves and its effect on Clostridium perfringens (anaerobe) growth
Osman ZA, et al.
European journal of molecular biotechnology, 3(4), 267-285 (2013)

Protocols

Enzymatic Assay of Ferredoxin NADP Reductase - To standardize a procedure for the enzymatic assay of Ferredoxin NADP Reductase (EC.1.18.12).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service