Skip to Content
Merck
All Photos(1)

Key Documents

F0382

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Rabbit IgG (whole molecule)–Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

indirect immunofluorescence: 1:80

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgGs are glycoprotein antibodies that modulate several immune responses. Rabbit IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rabbit IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins. Anti-Rabbit IgG (whole molecule)-FITC antibody reacts with rabbit serum and rabbit IgG.

Specificity

Binds all rabbit Igs.

Immunogen

Purified rabbit IgG

Application

Anti-Rabbit IgG (whole molecule)-FITC antibody is suitable for use in immunofluorescence .

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Sussman et al.
The Journal of clinical investigation, 101(1), 51-61 (1998-02-14)
Loss of myofibril organization is a common feature of chronic dilated and progressive cardiomyopathy. To study how the heart compensates for myofibril degeneration, transgenic mice were created that undergo progressive loss of myofibrils after birth. Myofibril degeneration was induced by
M J Laskowski
Planta, 181(1), 44-52 (1990-04-01)
The orientation of microtubules in cells of redlight-grown pea plants (Pisum sativum L.) was examined by means of immunofluorescence. Microtubules (MTs) in rapidly elongating, subepidermal cells commonly form multiple, parallel strands that run transverse to the cell's axis of elongation.
Claire Joly Condette et al.
PloS one, 9(7), e102217-e102217 (2014-07-16)
The epithelium's barrier function is crucial for maintaining homeostasis and preventing the passage of food antigens and luminal bacteria. This function is essentially subserved by tight junctions (TJs), multiprotein complexes located in the most apical part of the lateral membrane.
L K Zhuang et al.
Cell death & disease, 7, e2203-e2203 (2016-04-23)
MicroRNA (miRNA) and long non-coding RNA (lncRNA) have been demonstrated to participate in the progression of many cancers. Hepatocellular carcinoma (HCC) is one of the most common and aggressive malignant tumors worldwide, while the molecular mechanisms underlying HCC tumorigenesis are
Ga Bin Park et al.
Molecular medicine reports, 16(2), 1826-1836 (2017-06-20)
Cluster of differentiation (CD) 44 and epidermal growth factor (EGF) are closely involved in cellular migration and have been used as stem cell markers. Although the hyaluronan (HA)‑binding CD44 is responsible for enhanced cellular motility, the mechanism underlying its actions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service