Skip to Content
Merck
All Photos(1)

Key Documents

EHU155151

Sigma-Aldrich

MISSION® esiRNA

targeting human EP300

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATACCGAACCAAAGCCCTCTTTGCCTTTGAAGAAATTGATGGTGTTGACCTGTGCTTCTTTGGCATGCATGTTCAAGAGTATGGCTCTGACTGCCCTCCACCCAACCAGAGGAGAGTATACATATCTTACCTCGATAGTGTTCATTTCTTCCGTCCTAAATGCTTGAGGACTGCAGTCTATCATGAAATCCTAATTGGATATTTAGAATATGTCAAGAAATTAGGTTACACAACAGGGCATATTTGGGCATGTCCACCAAGTGAGGGAGATGATTATATCTTCCATTGCCATCCTCCTGACCAGAAGATACCCAAGCCCAAGCGACTGCAGGAATGGTACAAAAAAATGCTTGACAAGGCTGTATCAGAGCGTATTGTCCATGACTACAAGGATATTTTTAAACAAGCTACTGAAGATAGATTAACAAGTGCAAAGGAATTGCCTTATTTCGAGGGTGATTTCTGGCCCAATGTTCTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Tao et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 106, 1727-1733 (2018-08-19)
Pulmonary fibrosis is strongly correlated with inflammation factors, cytokine, and collagen secretion, whereby discoidin domain receptor 1 (DDR1) signaling plays an important role. EP300 is defined as an acetyltransferase that can acetylate histone and has been broadly studied in several
Alexander Ring et al.
BMC cancer, 20(1), 1076-1076 (2020-11-11)
Triple negative breast cancer (TNBC) is an aggressive breast cancer subtype with basal features, lacking the expression of receptors targeted successfully in other breast cancer subtypes. Treatment response to adjuvant and neoadjuvant chemotherapy is often short-lived and metastatic spread occurs
Mizuo Ando et al.
Nature communications, 10(1), 2188-2188 (2019-05-18)
Although promoter-associated CpG islands have been established as targets of DNA methylation changes in cancer, previous studies suggest that epigenetic dysregulation outside the promoter region may be more closely associated with transcriptional changes. Here we examine DNA methylation, chromatin marks
Karen Dendoncker et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(26), 12942-12951 (2019-06-12)
Glucocorticoid resistance (GCR) is defined as an unresponsiveness to the therapeutic effects, including the antiinflammatory ones of glucocorticoids (GCs) and their receptor, the glucocorticoid receptor (GR). It is a problem in the management of inflammatory diseases and can be congenital
Howard E Boudreau et al.
Oncotarget, 8(27), 44379-44397 (2017-06-03)
Previously, we showed wild-type (WT) and mutant (mut) p53 differentially regulate reactive oxygen species (ROS) generation by NADPH oxidase-4 (NOX4): p53-WT suppresses TGFβ-induced NOX4, ROS and cell migration, whereas tumor-associated mut-p53 proteins enhance NOX4 expression and cell migration. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service