Skip to Content
Merck
All Photos(1)

Key Documents

EHU071991

Sigma-Aldrich

MISSION® esiRNA

targeting human VCP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGAAGCCATCAATGAGGACAACAGTGTGGTGTCCTTGTCCCAGCCCAAGATGGATGAATTGCAGTTGTTCCGAGGTGACACAGTGTTGCTGAAAGGAAAGAAGAGACGAGAAGCTGTTTGCATCGTCCTTTCTGATGATACTTGTTCTGATGAGAAGATTCGGATGAATAGAGTTGTTCGGAATAACCTTCGTGTACGCCTAGGGGATGTCATCAGCATCCAGCCATGCCCTGATGTGAAGTACGGCAAACGTATCCATGTGCTGCCCATTGATGACACAGTGGAAGGCATTACTGGTAATCTCTTCGAGGTATACCTTAAGCCGTACTTCCTGGAAGCGTATCGACCCATCCGGAAAGGAGACATTTTTCTTGTCCGTGGTGGGATGCGTGCTGTGGAGTTCAAAGTGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sevil Cayli et al.
Theriogenology, 158, 196-206 (2020-09-24)
p97/valosin-containing protein (VCP) is expressed in many cells and plays critical functions in a broad range of diverse cellular processes. Because it is expressed in the mouse testes, predominantly in Sertoli cells, and is known to play a critical role
Wallaya Phongphaew et al.
Virus research, 228, 114-123 (2016-12-05)
Valosin-containing protein (VCP) is classified as a member of the type II AAA
Richard Wargachuk et al.
Cellular signalling, 30, 50-58 (2016-11-27)
GPCRs form signalling complexes with other receptors as part of dimers, G proteins and effector partners. A proteomic screen to identify proteins that associate with the β
Katrin Schweitzer et al.
Journal of cellular and molecular medicine, 20(1), 58-70 (2015-10-16)
Cullin-RING-ubiquitin-ligase (CRL)-dependent ubiquitination of the nuclear factor kappa B (NF-κB) inhibitor IκBα and its subsequent degradation by the proteasome usually precede NF-κB/RelA nuclear activity. Through removal of the CRL-activating modification of their cullin subunit with the ubiquitin (Ub)-like modifier NEDD8
Eva M van Well et al.
The EMBO journal, 38(9) (2019-03-20)
Neurodegenerative diseases are characterized by the accumulation of misfolded proteins in the brain. Insights into protein quality control mechanisms to prevent neuronal dysfunction and cell death are crucial in developing causal therapies. Here, we report that various disease-associated protein aggregates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service