Skip to Content
Merck
All Photos(3)

Key Documents

E0407

Sigma-Aldrich

Estramustine sodium phosphate

Synonym(s):

Estra-1,3,5(10)triene-3,17ß-diol 3-[N,N-bis(2-chloroethyl)carbamate] 17-(dihydrogen phosphate) sodium salt, Estradiol 3-bis(2-chloroethyl)carbamate 17-(dihydrogen phosphate) sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30Cl2NNa2O6P
CAS Number:
Molecular Weight:
564.35
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.28

Assay

≥85.0% (HPLC)

form

powder

solubility

H2O: 50 mg/mL

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@]12[C@@]([C@@](CC[C@@H]3OP([O-])([O-])=O)([H])[C@]3(C)CC2)([H])CCC4=C1C=CC(OC(N(CCCl)CCCl)=O)=C4.[Na+].[Na+]

InChI

1S/C23H32Cl2NO6P.2Na/c1-23-9-8-18-17-5-3-16(31-22(27)26(12-10-24)13-11-25)14-15(17)2-4-19(18)20(23)6-7-21(23)32-33(28,29)30;;/h3,5,14,18-21H,2,4,6-13H2,1H3,(H2,28,29,30);;/q;2*+1/p-2/t18-,19-,20+,21+,23+;;/m1../s1

InChI key

IIUMCNJTGSMNRO-VVSKJQCTSA-L

Gene Information

human ... ESR2(2100)

Biochem/physiol Actions

Estradiol mustard prodrug, which binds estradiol receptors irreversibly. Highly effective in treatment of advanced metastatic prostate cancer. Depolymerizes microtubules by binding to microtubule associated proteins (MAPs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service