Skip to Content
Merck
All Photos(1)

Key Documents

D8941

Sigma-Aldrich

2,6-Difluoro-4-[2-(phenylsul­fonyl­amino)­ethyl­thio]­phenoxy­acet­amide

≥98% (HPLC), solid

Synonym(s):

PEPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N2O4S2F2
CAS Number:
Molecular Weight:
402.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

white

solubility

DMSO: >20 mg/mL

SMILES string

NC(=O)COc1c(F)cc(SCCNS(=O)(=O)c2ccccc2)cc1F

InChI

1S/C16H16F2N2O4S2/c17-13-8-11(9-14(18)16(13)24-10-15(19)21)25-7-6-20-26(22,23)12-4-2-1-3-5-12/h1-5,8-9,20H,6-7,10H2,(H2,19,21)

InChI key

GTACSIONMHMRPD-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

2,6-Difluoro-4-[2-(phenylsul­fonyl­amino)­ethyl­thio]­phenoxy­acet­amide is a structurally novel, selective, high affinity AMPA ionotropic glutamate receptor agonist that reduces ischemia-induced performance deficits in rats.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weisheng Chen et al.
Neuroscience letters, 612, 126-131 (2015-12-08)
Infralimbic cortex (IL) is proposed to suppress cocaine seeking after extinction, but whether the IL regulates the extinction and reinstatement of heroin-seeking behavior is unknown. To address this issue, the male SD rats were trained to self-administer heroin under a
Amirbek Bekeshev et al.
Polymers, 12(7) (2020-07-02)
The conducted studies have proven the possibility of the directed control of operational properties of epoxy composites, due to the addition of finely-ground ocher into their composition, and the use of microwave modification of the epoxy composition. The rational content
Yanchao Xu et al.
Journal of colloid and interface science, 565, 23-34 (2020-01-14)
Nanofiltration (NF) membranes with tannic acid (TA) based selective layers have been intensively studied in recent years, but they still suffer from poor inorganic salt rejection. This study provided a first report on mussel-inspired co-deposition of TA and amine to
Quan-Guang Zhang et al.
Hippocampus, 20(1), 65-77 (2009-03-31)
alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptors (AMPARs) are responsible for excitotoxicity induced by ischemic injury in hippocampal CA1 neurons, whereas the molecular mechanisms responsible for their neurotrophic activities are much less studied. Here, we examined the neuroprotective effect of positive modeulation of AMPARs
Clarissa Perez Faria et al.
Scientific reports, 10(1), 6234-6234 (2020-04-12)
The protozoan Giardia lamblia is the most common cause of parasitic gastrointestinal infection worldwide. The parasite developed sophisticated, yet not completely disclosed, mechanisms to escape immune system and growth in the intestine. To further understand the interaction of G. lamblia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service