Skip to Content
Merck
All Photos(1)

Key Documents

D5891

Sigma-Aldrich

1,1-Dimethyl-4-phenylpiperazinium iodide

≥98% (TLC or titration)

Synonym(s):

DMPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19IN2
CAS Number:
Molecular Weight:
318.20
Beilstein:
3746109
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC or titration)

form

powder

mp

234-238 °C (lit.)

solubility

H2O: 21 mg/mL

storage temp.

−20°C

SMILES string

[I-].C[N+]1(C)CCN(CC1)c2ccccc2

InChI

1S/C12H19N2.HI/c1-14(2)10-8-13(9-11-14)12-6-4-3-5-7-12;/h3-7H,8-11H2,1-2H3;1H/q+1;/p-1

InChI key

XFZJGFIKQCCLGK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

1,1-Dimethyl-4-phenylpiperazinium iodide (DMPP) has been used as a non-selective nicotinic acetylcholine receptor (nAchR) agonist:
  • to study the roles of α7 and α4β2 nAchRs in autonomic regulation of cardiovascular responses in urethane-anesthetized mice
  • to study the mechanism involved in improving glucose tolerance in diet-induced obese (DIO) mice treated with DMPP
  • to study its pharmacological effect on enteric viscerofugal neurons in the myenteric plexus of guinea-pig colon

Biochem/physiol Actions

1,1-Dimethyl-4-phenylpiperazinium iodide (DMPP) is a non-selective nicotinic acetylcholine receptor (nAchR) agonist. It acts as a stimulating agent for sympathetic ganglia. DMPP exhibits specific action on ganglia and adrenal medullary tissue hence, it finds its application in the diagnosis of pheochromocytoma.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Hibberd et al.
Neuroscience, 275, 272-284 (2014-05-13)
Enteric viscerofugal neurons are mechanosensory interneurons that form the afferent limb of intestino-intestinal reflexes involving prevertebral sympathetic neurons. Fast synaptic inputs to viscerofugal neurons arise from other enteric neurons, but their sources are unknown. We aimed to describe the origins
D Manetti et al.
Bioorganic & medicinal chemistry, 7(3), 457-465 (1999-04-29)
A series of piperazine derivatives, obtained by hybridization of N1-acetyl-N4-dimethyl-piperazinium iodide (1, ADMP) and N1-phenyl-N4-dimethyl-piperazinium iodide (3, DMPP) or of the corresponding tertiary bases (2, 4) with arecoline (5) and arecolone (6) or by isosteric substitution of the phenyl ring
J J Galligan
The Journal of pharmacology and experimental therapeutics, 291(1), 92-98 (1999-09-22)
Nicotinic acetylcholine receptors (nAChRs) localized to excitatory longitudinal muscle motoneurons were studied in segments of guinea pig ileum maintained in vitro. Longitudinal muscle contractions caused by the nAChRs agonists, dimethylphenylpiperazinium (DMPP), nicotine, and cytisine were measured using isometric strain gauge
E S Vizi et al.
Brain research. Brain research reviews, 30(3), 219-235 (1999-11-24)
Neuronal nicotinic acetylcholine receptors (nAChRs) belong to a family of ligand-gated channels closely related to but distinct from the muscle nAChRs. Recent progress in neurochemical and pharmacological methods supports the hypothesis of presynaptically located nAChRs on axon terminals and indicates
Cardiovascular action of 1,1-dimethyl-4-phenylpiperazinium iodide (DMPP); a ganglion stimulating agent useful in the diagnosis of pheochromocytoma.
I H PAGE et al.
The American journal of medicine, 15(5), 675-683 (1953-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service