Skip to Content
Merck
All Photos(3)

Key Documents

D0633

Sigma-Aldrich

7-(2,3-Dihydroxypropyl)theophylline

Synonym(s):

7-(2,3-Dihydroxypropyl)-3,7-dihydro-1,3-dimethyl-1H-purine-2,6-dione, Diprophylline, Dyphylline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N4O4
CAS Number:
Molecular Weight:
254.24
Beilstein:
284563
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

form

powder

mp

161-162 °C (lit.)

SMILES string

CN1C(=O)N(C)c2ncn(CC(O)CO)c2C1=O

InChI

1S/C10H14N4O4/c1-12-8-7(9(17)13(2)10(12)18)14(5-11-8)3-6(16)4-15/h5-6,15-16H,3-4H2,1-2H3

InChI key

KSCFJBIXMNOVSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

7-(2,3-Dihydroxypropyl)theophylline is also called as dyphylline. Dyphylline serves as a bronchodilator and aids in better air flow through the lungs. It reduces the symptoms of chronic lung disorder such as asthma and bronchitis.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The AARP Guide to Pills: Essential Information on More Than 1,200 Prescription and Nonprescription Medicines, Including Generics, 302-302 (2006)
Mario Grassi et al.
International journal of pharmaceutics, 276(1-2), 107-114 (2004-04-29)
A mathematical model able to describe the release kinetics of two model drugs (Diprophylline and Theophylline) from partially coated hydroxypropylmethylcellulose (HPMC, Methocel) K4M) matrices is presented. As solvent interaction with the system and drug release can only take place in
Joshua Müller et al.
Journal of pharmaceutical and biomedical analysis, 53(4), 884-894 (2010-07-20)
A multivariate model was constructed by correlating Raman spectral data with coated amount of the API diprophylline using Partial Least Squares. In agreement with ICH guideline Q2 the method was validated in order to achieve the requirement of demonstrating that
Therapeutic Drug Monitoring and Toxicology by Liquid Chromatography (2017)
Sinan Güres et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 80(1), 122-129 (2011-10-20)
The aim of this study was to use a mechanistically realistic mathematical model based on Fick's second law to quantitatively predict the release profiles from solid lipid extrudates consisting of a ternary matrix. Diprophylline was studied as a freely water-soluble

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service