Skip to Content
Merck
All Photos(1)

Key Documents

1.2B4

10070103, human pancreas (islets), Epitheloid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

1.2B4, from human pancreas(islets), 10070103

biological source

human pancreas (islets)

growth mode

Adherent

karyotype

Modal chromosome number 67-71

morphology

Epitheloid

products

1.2B4 cells have been shown to express insulin, glucokinase and IAPP by immunocytochemistry. These cells express the GLUT1 glucose transporter.

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

Cell Line Description

1.2B4 is a hybrid cell line formed by the electrofusion of a primary culture of human pancreatic islets with HuP-T3, a human pancreatic carcinoma cell line (Sigma catalog number 93121055). 1.2B4 has been shown to be tumourigenic when transplanted into a SCID mouse host. The cell line has applications in the study of pancreatic cell biology. 1.2B4 cells provide a method of producing pure insulin secreting cells. This is an alternative to the use of primary tissue in cell transplantation therapies for type 1 diabetes.

DNA Profile

STR-PCR Data:
Amelogenin: X,Y
CSF1PO: 10
D13S317: 9,11
D16S539: 10,13
D5S818: 10,14
D7S820: 12
THO1: 9
TPOX: 8,12
vWA: 18

Culture Medium

RPMI-1640 + 2 mM Glutamine + 10% FCS / FBS

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 , i.e., seeding at 2-4 x 104 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37 °C. Population doubling approx 20hrs. At confluence 105 cells/cm2 can be expected.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Legal Information

Flatt P & McClenaghan N (2004) Insulin producing cell-line US patent number 20040005702 (A1). Also WO0210346 (A2).

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service