Skip to Content
Merck
All Photos(1)

Key Documents

C6356

Greiner culture flasks, tissue culture treated

T-25 flask, canted neck, surface area 25 cm2, capacity 50 mL, measuring grid, w/ standard cap

Synonym(s):

culture flask, tissue culture flask, tissue culture flasks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.13

material

canted neck
polystyrene flask
screw top red high-density polyethylene cap

sterility

sterile

feature

DNase free
RNase free
with cap
T-25 flask
measuring grid, w/ standard cap

packaging

case of 200 (20 bags x 10 flasks)

manufacturer/tradename

Greiner 690170

capacity

50 mL

surface area

25 cm2

working volume

10 mL

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

TC treated for maximum cell attachment.

Features and Benefits

  • Greiner culture flasks offer optimal cell adhesion through physical surface treatment.
  • Flasks are non-cytotoxic, non-pyrogenic and free of detectable DNase, RNase and human DNA.
  • The specific design facilitates optimal access to the samples with a cell scraper or pipette.
  • The canted neck ensures additional protection against contamination.
  • A standard screw cap with a secured click-in ventilation position enables gas exchange within the flasks.

NOTE: For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.

Legal Information


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service