Skip to Content
Merck
All Photos(1)

Key Documents

C4999

Sigma-Aldrich

Proinsulin C-Peptide (55-89) human

≥95% (HPLC)

Synonym(s):

Proinsulin connecting peptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C153H259N49O52
CAS Number:
Molecular Weight:
3616.99
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Proinsulin C-Peptide (55-89) human, bioactive peptide hormone

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H](NC(=O)[C@H](CCC(N)=O)NC(=O)CNC(=O)[C@@H](NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](C)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)CCCNC(N)=N)C(C)C)C(C)C)C(=O)NCC(=O)NCC(=O)NCC(=O)N1CCC[C@H]1C(=O)NCC(=O)N[C@@H](C)C(=O)NCC(=O)N[C@@H](CO)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N2CCC[C@H]2C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(O)=O)C(=O)NCC(=O)N[C@@H](CO)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C153H259N49O52/c1-71(2)55-94(193-134(237)91(40-48-119(222)223)189-148(251)122(78(15)16)200-135(238)86(33-41-105(156)205)179-112(212)67-175-147(250)121(77(13)14)199-136(239)88(35-43-107(158)207)188-139(242)97(58-74(7)8)194-142(245)100(61-120(224)225)197-133(236)90(39-47-118(220)221)182-124(227)80(18)177-129(232)89(38-46-117(218)219)186-130(233)84(29-23-51-167-152(162)163)183-126(229)82(155)27-22-50-166-151(160)161)128(231)172-63-110(210)169-62-109(209)170-68-115(215)201-53-25-31-103(201)145(248)174-64-111(211)176-79(17)123(226)171-65-113(213)180-101(69-203)144(247)196-99(60-76(11)12)141(244)190-92(36-44-108(159)208)149(252)202-54-26-32-104(202)146(249)198-95(56-72(3)4)137(240)178-81(19)125(228)192-96(57-73(5)6)138(241)185-85(37-45-116(216)217)127(230)173-66-114(214)181-102(70-204)143(246)195-98(59-75(9)10)140(243)187-87(34-42-106(157)206)132(235)184-83(28-20-21-49-154)131(234)191-93(150(253)254)30-24-52-168-153(164)165/h71-104,121-122,203-204H,20-70,154-155H2,1-19H3,(H2,156,205)(H2,157,206)(H2,158,207)(H2,159,208)(H,169,210)(H,170,209)(H,171,226)(H,172,231)(H,173,230)(H,174,248)(H,175,250)(H,176,211)(H,177,232)(H,178,240)(H,179,212)(H,180,213)(H,181,214)(H,182,227)(H,183,229)(H,184,235)(H,185,241)(H,186,233)(H,187,243)(H,188,242)(H,189,251)(H,190,244)(H,191,234)(H,192,228)(H,193,237)(H,194,245)(H,195,246)(H,196,247)(H,197,236)(H,198,249)(H,199,239)(H,200,238)(H,216,217)(H,218,219)(H,220,221)(H,222,223)(H,224,225)(H,253,254)(H4,160,161,166)(H4,162,163,167)(H4,164,165,168)/t79-,80-,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,121-,122-/m0/s1

InChI key

JEPNOJXFZHBCTB-ZCUALFGZSA-N

Amino Acid Sequence

Arg-Arg-Glu-Ala-Glu-Asp-Leu-Gln-Val-Gly-Gln-Val-Glu-Leu-Gly-Gly-Gly-Pro-Gly-Ala-Gly-Ser-Leu-Gln-Pro-Leu-Ala-Leu-Glu-Gly-Ser-Leu-Gln-Lys-Arg

General description

Human proinsulin, the single-chain peptide precursor of insulin, consists of the insulin A and B chains connected by the 31 amino acid C-peptide. Cleavage of proinsulin by proteolytic enzymes produces insulin and free C-peptide. The product here is the 35-mer peptide (Arg−Arg-C-Peptide-Lys-Arg) observed to separate the insulin A and B chains.

Biochem/physiol Actions

C-Peptide Fragment (55-89) is a fragment derived from the proinsulin C-peptide. C-peptide has cell signaling activity that involves calcium-dependent intracellular signaling. C-Peptide Fragment 55-89 may be used to help researchers understand the role of specific sequences within the C-peptide in binding, conformation and function of C-peptide.
C-Peptide was initially thought to have no intrinsic biological activity. More recent research has identified multiple biological effects of C-peptide, including binding to cell membranes, activation of signaling pathways, and stimulation of Na+,K+-ATPase and endothelial nitric oxide synthetase. However, the exact function of C-peptide is unclear due to conflicting physiological effects.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C-Peptide: A new mediator of atherosclerosis in diabetes.
Vasic, D., and Walcher, D.
Mediators of Inflammation, 858692-858692 (2012)
Proinsulin C-peptide prevents type-1 diabetes-induced decrease of renal Na+-K+-ATPase α1-subunit in rats.
Nordquist L, Shimada K, et al.
Diabetes and Metabolism, 26, 193-199 (2010)
R S Mughal et al.
Diabetologia, 53(8), 1761-1771 (2010-05-13)
Endothelial cells (ECs) and smooth muscle cells (SMCs) play key roles in the development of intimal hyperplasia in saphenous vein (SV) bypass grafts. In diabetic patients, insulin administration controls hyperglycaemia but cardiovascular complications remain. Insulin is synthesised as a pro-peptide
Intracellular signaling by C-peptide.
Hills, C.E. and Brunskill, N.J.
Experimental Diabetes Research, 635158-635158 (2008)
The conformational protential of porcine proinsulin C-peptide.
Vogt HP, Wollmer A, et al.
Hoppe-Seyler'S Zeitschrift fur Physiologische Chemie, 357, 107-116 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service