Skip to Content
Merck
All Photos(2)

Key Documents

C4490

Sigma-Aldrich

Anti-Caveolin-1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Caveolin-1, Anti-VIP21

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 22 kDa

species reactivity

human, rat, mouse

technique(s)

indirect immunofluorescence: 4-8 μg/mL using HUVEC cell line
indirect immunofluorescence: suitable
western blot: 1-2 μg/mL using whole cell extract of the human endothelial ECV304 cell line and the mouse fibroblast NIH3T3 cell line

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAV1(857)
mouse ... Cav1(12389)
rat ... Cav1(25404)

Looking for similar products? Visit Product Comparison Guide

General description

Caveolin, a 20-24 kDa integral transmembrane protein, has been identified as a principal component of caveolae membranes. Caveolin (also termed VIP21) exists in several isoforms termed caveolin-1, caveolin-2 and caveolin-3. Caveolin-1 (CAV1) (20- 22 kDa) can exist as two isoforms, caveolin-1α and -1β due to alternative splicing of the mRNA. Caveolin-1 and -2 have similar tissue distribution.

Specificity

The sequence is highly conserved in many species, e.g., identical in rat, mouse, bovine, and dog caveolin-1. The sequence is not found in caveolin-2 or caveolin-3.

Immunogen

synthetic peptide corresponding to a region at the N-terminus of human caveolin-1 (amino acids 2-20).

Application

Anti-Caveolin-1 antibody produced in rabbit has been used in:
  • immunoblotting
  • immunofluorescence
  • immunohistochemistry

Biochem/physiol Actions

Caveolin can simultaneously recognize GPI-linked proteins and interact directly with a number of caveolae associated downstream signaling molecules, such as H-Ras, hetero-trimeric G-proteins, annexin-II, epidermal growth factor receptor (EGFR), protein kinase C, src-family tyrosine kinases and nitric oxide synthase (NOS) isoforms. Caveolin-1 participates in angiogenesis and is expected to play a major role in the growth and evolution of cancer. It participates in glucose metabolism by controlling the expression of glucose transporters.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masashi Ogasawara et al.
Acta neuropathologica communications, 10(1), 176-176 (2022-12-09)
Oculopharyngodistal myopathy (OPDM) and oculopharyngeal muscular dystrophy (OPMD) are similar and even believed to be indistinguishable in terms of their myopathological features. To address the diagnostic gap, we evaluated the muscle biopsy samples for p62 expression by immunohistochemistry and compared
Brian C Russo et al.
mBio, 10(3) (2019-05-30)
Many Gram-negative bacterial pathogens require a type 3 secretion system (T3SS) to deliver effector proteins into eukaryotic cells. Contact of the tip complex of the T3SS with a target eukaryotic cell initiates secretion of the two bacterial proteins that assemble
Ingrid R Niesman et al.
Molecular and cellular neurosciences, 56, 283-297 (2013-07-16)
Microglia are ramified cells that serve as central nervous system (CNS) guardians, capable of proliferation, migration, and generation of inflammatory cytokines. In non-pathological states, these cells exhibit ramified morphology with processes intermingling with neurons and astrocytes. Under pathological conditions, they
Caveolin-1 is involved in reactive oxygen species-induced SHP-2 activation in astrocytes
Yun JH, et al.
Experimental & Molecular Medicine, 43(12), 660-660 (2011)
Study of caveolin-1 gene expression in whole adipose tissue and its subfractions and during differentiation of human adipocytes
Fernandez RJM, et al.
Nutrition & Metabolism, 7(1), 20-20 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service