Skip to Content
Merck
All Photos(3)

Key Documents

A9542

Sigma-Aldrich

Anti-Rat IgG (whole molecule)−Peroxidase antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Rabbit Anti-Rat IgG (whole molecule)−HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

peroxidase conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat

technique(s)

direct ELISA: 1:40,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:500
western blot (chemiluminescent): 1:5,000 using detecting rat primary antibody

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG (Immunoglobulin G) antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.
Immunoglobulins have two heavy chains and two light chains connected by disulfide bond. It is a glycoprotein which mainly helps in immune defense. IgG is a major class of immunoglobulin. Rat has five immunoglobulin classes- IgM, IgG, IgA, IgD and IgGE. IgG has four subclasses- IgG1, IgG2a, IgG2b and IgG2c.

Specificity

Anti-Rat IgG (whole molecule) antibody recognizes rat (immunoglobulin G) IgG and is determined by immunoelectrophoresis (IEP) versus normal rat serum and rat IgG prior to conjugation.

Immunogen

purified rat IgG

Application

Anti-Rat IgG (whole molecule) Peroxidase antibody produced in rabbit has been used in western blotting and enzyme linked immunosorbent assay (ELISA).

Biochem/physiol Actions

IgGs are glycoprotein antibodies that modulate several immune responses. Rat IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rat IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins.
Immunoglobulin G (IgG) participates in hypersensitivity type II and type III. It helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 0.05% MIT

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elicitors and defense gene induction in plants with altered lignin compositions
Gallego-GL, et al.
The New phytologist, 219, 1235-1251 (2018)
Mégane Nascimento et al.
Frontiers in immunology, 14, 1224383-1224383 (2023-12-26)
Chronic obstructive pulmonary disease (COPD) is a major health issue primarily caused by cigarette smoke (CS) and characterized by breathlessness and repeated airway inflammation. NLRP6 is a cytosolic innate receptor controlling intestinal inflammation and orchestrating the colonic host-microbial interface. However
Naringin protects myocardial cells from doxorubicin-induced apoptosis partially by inhibiting the p38MAPK pathway
Jian CY, et al.
Molecular Medicine Reports, 16(6), 9457-9463 (2017)
Lina Gallego-Giraldo et al.
The New phytologist, 219(4), 1235-1251 (2018-06-28)
A reduction in the lignin content in transgenic plants induces the ectopic expression of defense genes, but the importance of altered lignin composition in such phenomena remains unclear. Two Arabidopsis lines with similar lignin contents, but strikingly different lignin compositions
Adriana Trutzenberg et al.
Molecular plant pathology, 23(10), 1524-1537 (2022-07-19)
In barley (Hordeum vulgare), signalling rat sarcoma homolog (RHO) of plants guanosine triphosphate hydrolases (ROP GTPases) support the penetration success of Blumeria graminis f. sp. hordei but little is known about ROP activation. Guanine nucleotide exchange factors (GEFs) facilitate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service