Skip to Content
Merck
All Photos(1)

Key Documents

A1226

Sigma-Aldrich

Anti-Actopaxin antibody produced in rabbit

IgG fraction of antiserum, PBS solution

Synonym(s):

Anti-α-Parvin, Anti-CH-ILKBP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

PBS solution

mol wt

antigen 42 kDa

species reactivity

human, mouse

technique(s)

microarray: suitable
western blot: 1:3,000 using whole cell extract of the human endothelial ECV304 cell line.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PARVA(55742)
mouse ... Parva(57342)

Related Categories

General description

Actopaxin is a member of actin binding proteins that include beta-parvin/affixin and gamma-parvin. Actopaxin is known to associate with F-actin and paxillin LD motifs LD1 and LD4. This protein also directly binds to serine/threonine integrin-linked kinase (ILK). Studies have reported that actopaxin may be involved in integrin-dependent remodeling of the actin cytoskeleton during cell adhesion, motility and division.
Rabbit anti-actopaxin recognizes actopaxin (42 kDa). Staining of actopaxin in immunoblotting is specifically inhibited with actopaxin immunizing peptide (mouse, amino acids 35-53).

Immunogen

synthetic peptide corresponding to the N-terminal region of mouse actopaxin (amino acids 35-53). The sequence is identical in human actopaxin and has considerable homology (~70%) with β-parvin/affixin, but no homology with γ-parvin.

Application

Actopaxin was detected in HEK 293 cell lysates at 42 kDa by western blot using rabbit anti-actopaxin antibody at a 1:1000 dilution. In these cells, actopaxin associated with an ILK complex.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Rabbit anti-actopaxin can also be used for protein microaarays.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sotiris N Nikolopoulos et al.
The Journal of biological chemistry, 277(2), 1568-1575 (2001-11-06)
Paxillin is a focal adhesion adapter protein involved in integrin signaling. We have recently reported that the paxillin LD1 motif acts as a binding interface for both the actin-binding protein actopaxin and the serine/threonine integrin-linked kinase (ILK). In this report
Thitima Keskanokwong et al.
The Journal of biological chemistry, 282(32), 23205-23218 (2007-06-08)
Kidney anion exchanger 1 (kAE1) mediates chloride/bicarbonate exchange at the basolateral membrane of kidney alpha-intercalated cells, thereby facilitating bicarbonate reabsorption into the blood. Human kAE1 lacks the N-terminal 65 residues of the erythroid form (AE1, band 3), which are essential
S Yamaji et al.
The Journal of cell biology, 153(6), 1251-1264 (2001-06-13)
Focal adhesions (FAs) are essential structures for cell adhesion, migration, and morphogenesis. Integrin-linked kinase (ILK), which is capable of interacting with the cytoplasmic domain of beta1 integrin, seems to be a key component of FAs, but its exact role in
T M Olski et al.
Journal of cell science, 114(Pt 3), 525-538 (2001-02-15)
We have identified and cloned a novel 42-kDa protein termed alpha-parvin, which has a single alpha-actinin-like actin-binding domain. Unlike other members of the alpha-actinin superfamily, which are large multidomain proteins, alpha-parvin lacks a rod domain or any other C-terminal structural
Iveta Dobreva et al.
Journal of proteome research, 7(4), 1740-1749 (2008-03-11)
Protein-protein interactions play an essential role in the regulation of vital biological functions. Through a network of interactions, integrin-linked kinase (ILK) functions downstream of integrin receptors to control cell spreading, migration, growth, survival, and cell cycle progression. Despite many reports

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service