Skip to Content
Merck
All Photos(1)

Key Documents

87528

Sigma-Aldrich

Nα-Benzoyl-L-isoleucyl-L-glutamyl-glycyl-L-arginine-4-nitroanilide

≥97.0% (HPLC), powder

Synonym(s):

Bz-Ile-Glu-Gly-Arg-pNA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H43N9O9
CAS Number:
Molecular Weight:
697.74
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

Nα-Benzoyl-L-isoleucyl-L-glutamyl-glycyl-L-arginine-4-nitroanilide, ≥97.0% (HPLC)

Assay

≥97.0% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)c1ccccc1)C(=O)N[C@@H](CCC(O)=O)C(=O)NCC(=O)N[C@@H](CCCNC(N)=N)C(=O)Nc2ccc(cc2)[N+]([O-])=O

InChI

1S/C32H43N9O9/c1-3-19(2)27(40-28(45)20-8-5-4-6-9-20)31(48)39-24(15-16-26(43)44)29(46)36-18-25(42)38-23(10-7-17-35-32(33)34)30(47)37-21-11-13-22(14-12-21)41(49)50/h4-6,8-9,11-14,19,23-24,27H,3,7,10,15-18H2,1-2H3,(H,36,46)(H,37,47)(H,38,42)(H,39,48)(H,40,45)(H,43,44)(H4,33,34,35)/t19-,23-,24-,27-/m0/s1

InChI key

ZYECEUZMVSGTMR-LBDWYMBGSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Possible involvement of kinins in muscle energy metabolism.
G Dietze et al.
Advances in experimental medicine and biology, 167, 63-71 (1984-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service