Skip to Content
Merck
All Photos(1)

Key Documents

69152

Sigma-Aldrich

Disperse Blue 134

for microscopy

Synonym(s):

1,4-Bis(isopropylamino)anthraquinone, Solvent Blue 36

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N2O2
CAS Number:
Molecular Weight:
322.40
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy

form

powder

mp

170-172 °C

solubility

toluene: 0.01 g/10 mL, blue to very deep blue

εmax

≥400 at 587-597 nm in ethanol
≥550 at 635-645 nm in ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC(C)Nc1ccc(NC(C)C)c2C(=O)c3ccccc3C(=O)c12

InChI

1S/C20H22N2O2/c1-11(2)21-15-9-10-16(22-12(3)4)18-17(15)19(23)13-7-5-6-8-14(13)20(18)24/h5-12,21-22H,1-4H3

InChI key

BLFZMXOCPASACY-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Patrick Mulqueen
Advances in colloid and interface science, 106, 83-107 (2003-12-16)
This review will describe some of the changes occurring in formulation types employed and the further trends that are driving technologies. These will be illustrated by examples of water-based dispersion formulation technology for oil-in-water emulsions (EW) and suspension-emulsions (SE) as
T E Creekmore et al.
Journal of wildlife diseases, 30(4), 497-505 (1994-10-01)
A field study was conducted on Antigua, West Indies, to determine the feasibility of delivering an oral rabies vaccine or population control agent to free-ranging mongooses (Herpestes javanicus). Two biomarkers (tetracycline hydrochloride [THCL] and DuPont Oil Blue A dye) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service