Skip to Content
Merck
All Photos(1)

Key Documents

6902-50A

Sigma-Aldrich

Human Peripheral Blood T Cells: HPBT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

human peripheral blood

form

liquid

packaging

pkg of 5,000,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Suspension

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Human Peripheral Blood T Cells (HPBT) provide a useful in vitro tool for studying the biology and pathology of CD4+ cells. HPBT cells play critical roles in the regulation of immune responses, and are responsible for mediating many of the effector mechanisms of the immune system. Increased activity in the field of immunotherapy, coupled with the expense of clinical trials, has led to renewed interest in obtaining high purity human T cells to meet needs for research and development. Our CD4+ T Cells are isolated from human blood of healthy donors and cryopreserved immediately after isolation.

Cell Line Origin

Blood

Application

Pathology, cancer, autoimmune disorders, infectious diseases. Immune response regulation, direct destruction of infected cells, immunocompetence, scanning of intracellular environment, effector mechanisms, realistic in vitro model, immunotherapeutics. T cell origination, development, differentiation, expansion, proliferation, maturation and activation. Hematopoesis, stem cells, positive & negative selection. T cell receptors (TCRs), antigen recognition, costimulatory molecules, interaction with foreign antigens, signaling cascades and cytokine production.

Packaging

Immediately after isolation, 5,000,000 cells in DMEM containing 45% FBS & 10% DMSO.

Components

DMEM containing 45% FBS & 10% DMSO

Quality

Each lot was tested for proper morphology, CD4 positive selection, Negative for HIV, Hepatitis B, Hepatitis C

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service