Skip to Content
Merck
All Photos(1)

Key Documents

51441C

SAFC

Dulbecco′s Modified Eagle′s Medium/High Modified

liquid, sterile-filtered, with 4500 mg/L dextrose, with 4.0 mM L-glutamine, without sodium pyruvate, suitable for cell culture

Synonym(s):

Dulbecco’s Modified Eagle’s Medium - high glucose, DME, DMEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

components

glucose: high
phenol red: yes
sodium pyruvate: no
L-glutamine: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains four-fold concentrations of the amino acids and vitamins. The original formulation contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. Since then, it has been modified in several ways to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Each of these media offers a different combination of L-glutamine and sodium pyruvate. Additionally, the glucose levels have been raised to 4500 mg/L, contributing to the name "DMEM/High".

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H B Shi et al.
Journal of dairy science, 103(10), 9096-9101 (2020-08-24)
Proteasomes play a widespread role in the control of protein abundance via degrading ubiquitinated proteins. Activity of proteasomes is regulated by constitutive ATPases that respond to intracellular concentrations of ATP. Although recent data suggest a role of proteasomes in fatty
Daniela Fignani et al.
Frontiers in endocrinology, 11, 596898-596898 (2020-12-08)
Increasing evidence demonstrated that the expression of Angiotensin I-Converting Enzyme type 2 (ACE2) is a necessary step for SARS-CoV-2 infection permissiveness. In light of the recent data highlighting an association between COVID-19 and diabetes, a detailed analysis aimed at evaluating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service