Skip to Content
Merck
All Photos(1)

Key Documents

51412C

SAFC

Minimum Essential Medium

with Earle′s Balanced Salts, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Pharma Manufacturing

Synonym(s):

Minimum Essential Medium Eagle, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

description

for research or for further manufacturing use

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable
cell culture | stem cell: suitable

components

L-glutamine: no
NaHCO3: 2200 mg/L
phenol red: 10.62 mg/L
Earle’s salts (5% CO2): yes

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Minimum Essential Medium (MEM) with Earle′s Balanced Salts was developed by Harry Eagle as a result of his studies to determine the essential nutritional requirements of mouse L cells and HeLa cells in culture. He demonstrated that these cells require 13 amino acids and seven vitamins to grow and reproduce in vitro. MEM is a modification of Eagle′s earlier medium Basal Medium Eagle (BME), containing higher concentrations of the essential nutrients. This medium has demonstrated the ability to support a variety of normal and transformed cells in culture and contains Earle′s Balanced Salts, which makes it suitable for use in atmospheres charged with CO2 gas.

Application

Minimum essential medium (MEM) has been used as a growth medium for bone marrow mesenchymal stem cells (BMSCs) and neurons.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel pulsed STED microscopy method using FastFLIM and the phasor plots.
Sun Y, et al.
Proceedings of SPIE (2017)
Involvement of microRNA-23b in TNF-?-reduced BMSC osteogenic differentiation via targeting runx2.
Deng L, et al.
Journal of Bone and Mineral Metabolism, 1-13 (2017)
Debora Soncini et al.
Blood advances, 4(18), 4312-4326 (2020-09-12)
Metabolic reprogramming is emerging as a cancer vulnerability that could be therapeutically exploitable using different approaches, including amino acid depletion for those tumors that rely on exogenous amino acids for their maintenance. ʟ-Asparaginase (ASNase) has contributed to a significant improvement
Agata Nowacka et al.
Scientific reports, 10(1), 2054-2054 (2020-02-08)
PSD-95 is a major scaffolding protein of the post-synaptic density (PSD) of a glutamatergic synapse. PSD-95, via interactions with stargazin, anchors AMPA receptors at the synapse and regulates AMPAR currents. The expression of PSD-95 is regulated during synaptic plasticity. It
Robert Nason et al.
Journal of the Association for Research in Otolaryngology : JARO, 10(2), 151-160 (2009-01-16)
Osteoclasts are the only cells capable of carrying out bone resorption and therefore are responsible for the osteolysis seen in infectious diseases such as chronic otitis media and infected cholesteatoma. Pseudomonas aeruginosa is the most common organism isolated from these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service