Skip to Content
Merck
All Photos(1)

Key Documents

5123

Sigma-Aldrich

CD276 human

recombinant, expressed in E. coli, 0.5 mg protein/mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.75

biological source

human

recombinant

expressed in E. coli

description

0.1 mg recombinant human CD276 in 20 mM Tris-HCl buffer, containing NaCl, KCl, EDTA, L-arginine, DTT and glycerol.

sterility

Filtered sterilized solution

Assay

≥90% (SDS-PAGE)

form

liquid

packaging

pkg of 100 μg

concentration

0.5 mg protein/mL

technique(s)

cell culture | mammalian: suitable

accession no.

NP_001019907

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CD276(80381)

Application

Coating a plate well (6 well plate) with this recombinant CD276 protein in T cell specific medium at 1-10 μg/well allows for use 1) for human T cell / receptor interaction study in vitro or 2) as a highly purified recombinant antigen as cancer biomarker for diagnosis application development.

Use this procedure as a guideline to determine optimal coating conditions for the culture system of choice.
1. Thaw CD276 and dilute to desired concentration using serum-free medium or PBS. The final solution should be sufficiently dilute so the volume added covers the surface evenly (1-10 μg/well, 6 well plate).
Note: Use 1 ml PBS per well in a 6-well plate.
2. Add 1 - 10 μg protein to each well and incubate at 2 to 10 °C overnight.
3. After incubation, aspirate remaining material.
4. Plates are ready for use. They may also be stored at 2-8 °C damp or air dried if sterility is maintained.

Sequence

MASMTGGQQMGRGHHHHHHGNLYFQGEVQVPEDPVVALVGTDATLCCSFSPEPGFSLAQLNLIWQLTDTKQLVHSFAEGQDQGSAYANRTALFPDLLAQGNASLRLQRVRVADEGSFTCFVSIRDFGSAAVSLQVAAPYSKPSMTLEPNKDLRPGDTVTITCSSYQGYPEAEVFWQDGQGVPLTGNVTTSQMANEQGLFDVHSILRVVLGANGTYSCLVRNPVLQQDAHSSVTITPQRSPTGAVEVQVPEDPVVALVGTDATLRCSFSPEPGFSLAQLNLIWQLTDTKQLVHSFTEGRDQGSAYANRTALFPDLLAQGNASLRLQRVRVADEGSFTCFVSIRDFGSAAVSLQVAAPYSKPSMTLEPNKDLRPGDTVTITCSSYRGYPEAEVFWQDGQGVPLTGNVTTSQMANEQGLFDVHSVLRVVLGANGTYSCLVRNPVLQQDAHGSVTITGQPMTFPPEA

Preparation Note

The full-length extracellular domain of the human CD276 gene (29 - 466 aa) was constructed with 29 N-terminal T7/HIS-tag and expressed in E. coli as inclusion bodies. The final product was refolded using a unique “temperature shift inclusion body refolding” technology and chromatographically purified.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luana Calabrò et al.
Journal of cellular physiology, 226(10), 2595-2600 (2011-07-28)
No treatment prolongs the survival of malignant mesothelioma (MM) patients. Since MM elicits anti-tumor host's immune responses, immunotherapy represents a promising strategy for its control. Immunomodulatory antibodies against components of the B7 family of immunomodulatory molecules that regulate T cell
Zongliang Zhang et al.
Molecular therapy oncolytics, 17, 180-189 (2020-04-30)
Recently, B7-H3 was frequently reported to be overexpressed in various cancer types and has been suggested to be a promising target for cancer immunotherapy. In the present study, we analyzed the mRNA expression of B7-H3 in The Cancer Genome Atlas
A I Chapoval et al.
Nature immunology, 2(3), 269-274 (2001-02-27)
We describe here a newly identified member of the human B7 family, designated B7 homolog 3 (B7-H3), that shares 20-27% amino acid identity with other B7 family members. B7-H3 mRNA is not detectable in peripheral blood mononuclear cells, although it
Vibeke A Ingebrigtsen et al.
BMC cancer, 14, 602-602 (2014-08-21)
We have previously reported overexpression of the immunoregulatory protein B7-H3 in colorectal cancer and that nuclear expression predicted poor outcome in colon cancer patients. The present study was performed to examine the prognostic role of B7-H3 in an independent colorectal
Bo Gong et al.
The Journal of experimental medicine, 216(4), 982-1000 (2019-03-16)
Immune checkpoint blockade against programmed cell death 1 (PD-1) and its ligand PD-L1 often induces durable tumor responses in various cancers, including non-small cell lung cancer (NSCLC). However, therapeutic resistance is increasingly observed, and the mechanisms underlying anti-PD-L1 (aPD-L1) antibody

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service