Skip to Content
Merck
All Photos(1)

Key Documents

45199

Sigma-Aldrich

Enterolactone

~95% (HPLC)

Synonym(s):

trans-α,β-Bis(3-hydroxybenzyl)butyrolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18O4
CAS Number:
Molecular Weight:
298.33
Beilstein:
4699604
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.77

Assay

~95% (HPLC)

form

solid

storage temp.

2-8°C

SMILES string

Oc1cccc(C[C@@H]2COC(=O)[C@H]2Cc3cccc(O)c3)c1

InChI

1S/C18H18O4/c19-15-5-1-3-12(8-15)7-14-11-22-18(21)17(14)10-13-4-2-6-16(20)9-13/h1-6,8-9,14,17,19-20H,7,10-11H2/t14-,17+/m1/s1

InChI key

HVDGDHBAMCBBLR-PBHICJAKSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Enterolactone is the major human metabolite of dietary sesamin (lignan from sesame seeds). A phytoestrogen, it shows an inverse correlation with risk of breast cancer in one study.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Anne Tuomisto et al.
Scientific reports, 9(1), 11209-11209 (2019-08-03)
The dietary lignan metabolite, enterolactone, has been suggested to have anti-cancer functions, and high serum enterolactone concentrations have been associated with decreased risk of breast and prostate cancers. We hypothesized that serum enterolactone concentrations as a marker of plant-based foods
Mark J McCann et al.
Molecular nutrition & food research, 57(2), 212-224 (2012-11-14)
There is evidence that a mammalian lignan, enterolactone (ENL), decreases the proliferation rate of prostate cancer cells, although previous studies have used concentrations difficult to achieve through dietary modification. We have therefore investigated the anti-proliferative effects of ENL in an
Kirstine S Krogholm et al.
The British journal of nutrition, 108(10), 1904-1912 (2012-03-29)
Since collection of 24 h urine samples is very time consuming and difficult to obtain, epidemiological studies typically only obtain spot urine samples. The aim of the present study was to evaluate whether flavonoids and enterolactone in overnight urine could
Karin Preindl et al.
Analytical chemistry, 91(17), 11334-11342 (2019-08-10)
We are constantly exposed to a variety of environmental contaminants and hormones, including those mimicking endogenous estrogens. These highly heterogeneous molecules are collectively referred to as xenoestrogens and hold the potential to affect and alter the delicate hormonal balance of
Clare Whitton et al.
Public health nutrition, 21(15), 2735-2743 (2018-08-08)
Short screeners for assessing dietary quality are lacking in Asia. We recently developed a short thirty-seven-item diet screener (DS). The present study aimed to evaluate reproducibility and relative validity of the DS in assessing a priori dietary quality indices (DQI;

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service