Skip to Content
Merck
All Photos(1)

Key Documents

43794

Sigma-Aldrich

1,4-Dithioerythritol

≥99.0% (RT), BioUltra

Synonym(s):

erythro-1,4-Dimercapto-2,3-butanediol, erythro-2,3-Dihydroxy-1,4-butanedithiol, Cleland’s reagent, DTE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(OH)CH(OH)CH2SH
CAS Number:
Molecular Weight:
154.25
Beilstein:
1719756
EC Number:
MDL number:
UNSPSC Code:
12161504
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Assay

≥99.0% (RT)

form

powder

reaction suitability

reagent type: reductant

impurities

insoluble matter, passes filter test
≤0.5% 4,5-dihydroxy-1,2-dithiane

pH

4.0-6.0 (25 °C, 0.1 M in H2O)

mp

82-84 °C (lit.)
82-85 °C

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in H2O

UV absorption

λ: 260 nm Amax: 0.400
λ: 280 nm Amax: 0.050

storage temp.

2-8°C

SMILES string

O[C@@H](CS)[C@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4+

InChI key

VHJLVAABSRFDPM-ZXZARUISSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for maintaining −SH groups in the reduced state; quantitatively reduces disulfides.

Other Notes

Cleavage of disulfide bonds in proteins; Prevents oxidation of sulfhydryl-containing proteins during SDS-polyacrylamide gel electrophoresis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A simple method for stabilizing protein-sulfhydryl groups during SDS-gel electrophoresis.
L C Lane
Analytical biochemistry, 86(2), 655-664 (1978-06-01)
A specific and sensitive assay for disulfides.
W L Zahler et al.
The Journal of biological chemistry, 243(4), 716-719 (1968-02-25)
Reductive cleavage of cystine disulfides with tributylphosphine.
U T Rüegg et al.
Methods in enzymology, 47, 111-116 (1977-01-01)
Yang-Rae Kim et al.
ACS applied materials & interfaces, 2(1), 292-295 (2010-04-02)
A strategy for the rational design of a novel colorimetric sensor based on dithioerythritol-modified gold nanoparticles for the selective recognition of Hg2+ in aqueous media is presented. This approach relies on the combination of gold nanoparticles with Hg2+ through sulfur-Hg2+-sulfur
András Kolozsi et al.
Inorganic chemistry, 47(9), 3832-3840 (2008-04-03)
The aqueous solutions of arsenous acid with the meso and racemic forms of 1,4-dithiol-butane-2,3-diol, namely, dithioerythritol (dte) and dithiothreitol (dtt), respectively, were titrated pH-metrically in different molar ratios. The p K a values determined for As(OH) 3, and dtt were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service