Skip to Content
Merck
All Photos(1)

Key Documents

355A-1

Sigma-Aldrich

GLUT1 Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (355A-14)
vial of 0.5 mL concentrate (355A-15)
bottle of 1.0 mL predilute (355A-17)
vial of 1.0 mL concentrate (355A-16)
bottle of 7.0 mL predilute (355A-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

control

colorectal carcinoma, mesothelioma

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous

Gene Information

human ... SLC2A1(6513)

General description

Glucose transporter type I (GLUT1), a prototype member of GLUT superfamily, is a membrane-associated erythrocyte glucose transport protein. It is a major glucose transporter in the mammalian blood-brain barrier, and also mediates glucose transport in endothelial cells of the vasculature, adipose tissue, and cardiac muscle. GLUT1 is detectable in many human tissues including those of colon, lung, stomach, esophagus, and breast. GLUT1 is overexpressed in malignant cells and in a variety of tumors that include the breast, pancreas, cervix, endometrium, lung, mesothelium, colon, bladder, thyroid, bone, soft tissues, and oral cavity. Immunohistochemical detection of GLUT1 has been shown to discriminate between reactive mesothelium and malignant mesothelioma in more than one study.Anti-GLUT1 is also useful in distinguishing benign endometrial hyperplasia from atypical endometrial hyperplasia and adenocarcinoma.

Quality


IVD

IVD

IVD

RUO

Linkage

GLUT1 Positive Control Slides, Product No. 355S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Value of immunohistochemical markers in differentiating benign from malignant mesothelial lesions
Acurio A, et al.
United States and Canadian Academy of Pathology Annual Meeting, 2008 null
Alaa Afify et al.
Acta cytologica, 49(6), 621-626 (2006-02-03)
To evaluate the extent to which adenocarcinomas in body cavity fluids express GLUT-1 in comparison to currently available markers for adenocarcinomas. Archival paraffin-embedded cell blocks of serous fluids from 25 cases of benign effusions containing reactive mesothelial cells and 39
Yasufumi Kato et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 20(2), 215-220 (2006-12-29)
The separation of benign reactive mesothelium (RM) from malignant mesothelial proliferation can be a major challenge. A number of markers have been proposed, including epithelial membrane antigen, p53 protein, and P-glycoprotein. To date, however, no immunohistochemical marker that allows unequivocal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service