Skip to Content
Merck
All Photos(1)

Key Documents

30089

Sigma-Aldrich

L-Cysteine

≥98.5% (RT), suitable for HPLC, BioUltra

Synonym(s):

(R)-2-Amino-3-mercaptopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
121.16
Beilstein:
1721408
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

L-Cysteine, BioUltra, ≥98.5% (RT)

product line

BioUltra

Assay

≥98.5% (RT)

form

powder or crystals

optical activity

[α]20/D +8.0±0.5°, c = 5% in 5 M HCl

technique(s)

HPLC: suitable

impurities

insoluble matter, passes filter test
≤0.5% foreign amino acids

ign. residue

≤0.05% (as SO4)

loss

≤0.05% loss on drying, 20 °C (HV)

color

white

mp

240 °C (dec.) (lit.)

solubility

1 M HCl: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤500 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in 1 M HCl

UV absorption

λ: 260 nm Amax: 1.5
λ: 280 nm Amax: 0.2

SMILES string

N[C@@H](CS)C(O)=O

InChI

1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)/t2-/m0/s1

InChI key

XUJNEKJLAYXESH-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Cysteine has been used in quenching of unreacted maleimide during antibody grafting. It has also been used in the preparation of nitrosocysteine stock solution, that has been employed to induce S-nitrosylation to achieve reversible cysteine modification.

Biochem/physiol Actions

Cysteine is one of the functional amino acids that regulates metabolism for growth, reproduction, maintenance and immunity. Cysteine provides the disulfide linkage in proteins. It is directly associated with the transport of sulfur. Taurine, a metabolic product of cysteine acts as an antioxidant and controls the cellular redox state. Cysteine is involved in the formation of hydrogen sulfide that is utilized as a signaling molecule.
NMDA glutamatergic receptor agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of polyion complex micelles from poly(ethylene glycol)-block-polyions
Nuria Bayo-Puxan
Journal of Controlled Release : Official Journal of the Controlled Release Society, 156(2), 118-127 (2011)
Resin-assisted enrichment of thiols as a general strategy for proteomic profiling of cysteine-based reversible modifications
Jia Guo
Nature Protocols, 9(1), 64-75 (2014)
Amino acids: metabolism, functions, and nutrition
Guoyao Wu
Amino Acids, 37(1), 1-17 (2009)
Takeshi Annoura et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(5), 2158-2170 (2014-02-11)
The 10 Plasmodium 6-Cys proteins have critical roles throughout parasite development and are targets for antimalaria vaccination strategies. We analyzed the conserved 6-cysteine domain of this family and show that only the last 4 positionally conserved cysteine residues are diagnostic
Nicholas Kwiatkowski et al.
Nature, 511(7511), 616-620 (2014-07-22)
Tumour oncogenes include transcription factors that co-opt the general transcriptional machinery to sustain the oncogenic state, but direct pharmacological inhibition of transcription factors has so far proven difficult. However, the transcriptional machinery contains various enzymatic cofactors that can be targeted

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service