Skip to Content
Merck
All Photos(1)

Key Documents

156M-8

Sigma-Aldrich

CD56 (123C3.D5) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

123C3.D5, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (156M-84)
vial of 0.5 mL concentrate (156M-85)
bottle of 1.0 mL predilute (156M-87)
vial of 1.0 mL concentrate (156M-86)
bottle of 7.0 mL predilute (156M-88)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1κ

control

neuroblastoma

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous

Gene Information

human ... NCAM1(4684)

General description

Anti-CD56 recognizes two proteins of the neural cell adhesion molecule, the basic molecule expressed on most neuroectodermally derived cell lines, tissues and neoplasms (e.g. retinoblastoma, medulloblastomas, astrocytomas, neuroblastomas, and small cell carcinomas). It is also expressed on some mesodermally derived tumors (rhabdomyosarcoma).

Linkage

CD56 Positive Control Slides, Product No. 156S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Gerardy-Schahn et al.
International journal of cancer. Supplement = Journal international du cancer. Supplement, 8, 38-42 (1994-01-01)
Monoclonal antibodies (MAbs) ranked together as small-cell-lung-cancer (SCLC) Cluster I MAbs are directed against the neural cell adhesion molecule NCAM (CD 56) and have been shown to be useful reagents in SCLC diagnosis and therapy. We analyzed the epitopes recognized
R Michalides et al.
International journal of cancer. Supplement = Journal international du cancer. Supplement, 8, 34-37 (1994-01-01)
The presence of the neural cell adhesion molecule, NCAM, is indicative for a poor prognosis in lung-cancer patients. Using MAb 735, we have investigated the expression of polysialic acid, PSA, on NCAM in a spectrum of neuro-endocrine lung tumors, ranging
Jianguo Tao et al.
The American journal of surgical pathology, 26(1), 111-118 (2002-01-05)
We report an unusual case of aggressive natural killer (NK)-like cytotoxic T-cell lymphoma in a previously healthy immunocompetent West African male. He presented with a fever of unknown origin, subsequently developed erythematous skin nodules, generalized lymphadenopathy, and hepatosplenomegaly, and then
R E Kibbelaar et al.
European journal of cancer (Oxford, England : 1990), 27(4), 431-435 (1991-01-01)
We investigated the expression of the neural cell adhesion molecule (NCAM) in a series of surgically resected lung carcinomas of various histological subtypes by means of a panel of monoclonal antibodies recognising different N-CAM epitopes. In a subgroup of 56
Scott A Ely et al.
The American journal of pathology, 160(4), 1293-1299 (2002-04-12)
Unlike monoclonal gammopathy of undetermined significance (MGUS) or non-Hodgkin's lymphomas (NHLs) with plasmacytoid differentiation, multiple myeloma (MM) is commonly associated with lytic bone lesions. Although the mechanisms of increased osteoclast activity are partially understood, comparatively little is known about the

Related Content

Trusted IVDR-ready supplier of high-quality Cell Marque™ antibodies for diagnostic pathology and IHC, ensuring reliability and expert support.

Trusted IVDR-ready supplier of high-quality Cell Marque™ antibodies for diagnostic pathology and IHC, ensuring reliability and expert support.

Trusted IVDR-ready supplier of high-quality Cell Marque™ antibodies for diagnostic pathology and IHC, ensuring reliability and expert support.

Trusted IVDR-ready supplier of high-quality Cell Marque™ antibodies for diagnostic pathology and IHC, ensuring reliability and expert support.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service