Skip to Content
Merck
All Photos(1)

Key Documents

D5509

Sigma-Aldrich

Diatomaceous earth non-washed

powder

Synonym(s):

Diatomaceous earth, Diatomaceous silica, Kieselguhr

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:
NACRES:
NA.21

form

powder

color

white to light gray

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Highly insoluble, inert material for use as a filtration aid.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Messaouda Safa et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 65(10), 1729-1737 (2012-05-02)
The adsorption of Cu(2+), Zn(2+), Cd(2+) and Pb(2+) ions from aqueous solution by Algerian raw diatomite was studied. The influences of different sorption parameters such as contact pH solution, contact time and initial metal ions concentration were studied to optimize
Goran G Andrić et al.
Journal of economic entomology, 105(2), 670-678 (2012-05-23)
Insecticidal potential of natural zeolites and diatomaceous earths originating from Serbia against Sitophilus oryzae (L.) and Tribolium castaneum (Herbst) was evaluated. Two natural zeolite formulations (NZ and NZ Modified) were applied to wheat at rates of 0.50, 0.75, and 1.0
Manpreet Bariana et al.
International journal of pharmaceutics, 443(1-2), 230-241 (2013-01-05)
Diatomaceous earth (DE), or diatomite silica microparticles originated from fossilized diatoms are a potential substitute for its silica-based synthetic counterparts to address limitations in conventional drug delivery. This study presents the impact of engineered surface chemistry of DE microparticles on
Heezoo Kim et al.
Anesthesia and analgesia, 115(2), 244-252 (2012-05-16)
The activated clotting time (ACT) is widely used for monitoring heparin anticoagulation during cardiac surgery. Celite-based ACT values are prolonged when aprotinin is administered. MDCO-2010, a novel serine protease inhibitor, is currently being evaluated as a possible alternative to aprotinin.
Tino Jaschinski et al.
Rapid communications in mass spectrometry : RCM, 27(1), 109-116 (2012-12-15)
Organic matrices are the state-of-the-art ionization mediators in Laser Desorption/Ionization Mass Spectrometry (LDI-MS). Despite improvements in understanding matrix chemistry, interfering matrix-related signals complicate the analysis. Surface-assisted LDI techniques like desorption/ionization on silicon (DIOS) or nanostructure initiator mass spectrometry (NIMS) provide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service