Skip to Content
Merck
All Photos(1)

Key Documents

60710

Sigma-Aldrich

Kerosene

purum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
15101502
NACRES:
NA.21

vapor density

4.5 (vs air)

vapor pressure

0.23 mmHg ( 20 °C)

grade

purum

form

liquid

autoignition temp.

442 °F

expl. lim.

5 %

refractive index

n20/D 1.436

bp

190-250 °C (lit.)

density

0.8 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

General description

Kerosene is a medium petroleum fraction. It is widely used as commercial fuel. A kinetic reaction mechanism for the oxidation of kerosene has been investigated. A fuel processing system has been developed for improving the reforming activity of kerosene for use in polymer electrolyte fuel cells (PEFCs).

Application

Kerosene may be employed as liquid fuel for the stationary fuel cells.

Other Notes

Sales restrictions may apply

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 432-432 (1994)
Development of kerosene fuel processing system for PEFC.
Fukunaga T, et al.
Catalysis Today, 84(3), 197-200 (2003)
On the kinetics of hydrocarbons oxidation from natural gas to kerosene and diesel fuel.
Dagaut P.
Physical Chemistry Chemical Physics, 4(11), 2079-2094 (2002)
Jane Jose Vattathara et al.
Scientific reports, 10(1), 3790-3790 (2020-03-04)
The concept of substrate inhibition to prevent its phosphorylation has potential in drug discovery and is envisioned to treat the autoimmune disorder multiple sclerosis (MS). Glia maturation factor-β (GMF-β) Ser83 phosphorylation by protein kinase A (PKA) is pivotal in the
J A Namocatcat et al.
Journal of contaminant hydrology, 67(1-4), 177-194 (2003-11-11)
Evolution of trimethylbenzoic acids in the KC-135 aquifer at the former Wurtsmith Air Force Base (WAFB), Oscoda, MI was examined to determine the functionality of trimethylbenzoic acids as key metabolite signatures in the biogeochemical evolution of an aquifer contaminated with

Protocols

Summary application report for analysis of moisture in Kerosene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service