Skip to Content
Merck
All Photos(1)

Key Documents

59070

Sigma-Aldrich

2-Methylbutane

puriss. p.a., ≥99.5% (GC)

Synonym(s):

Isopentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)2
CAS Number:
Molecular Weight:
72.15
Beilstein:
1730723
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.07

vapor density

2.6 (vs air)

vapor pressure

11.17 psi ( 20 °C)

grade

puriss. p.a.

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

788 °F

expl. lim.

8.3 %

impurities

≤0.05% water

evapn. residue

≤0.001%

refractive index

n20/D 1.354 (lit.)
n20/D 1.354

bp

30 °C (lit.)

mp

-160 °C

density

0.62 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

storage temp.

2-8°C

SMILES string

CCC(C)C

InChI

1S/C5H12/c1-4-5(2)3/h5H,4H2,1-3H3

InChI key

QWTDNUCVQCZILF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methylbutane (Isopentane) is a branched chain alkane. It undergoes bromination at 75°C to afford 2-bromo-2-methyl butane. Its Raman spectra in vapor and solid phases at low temperatures (-196°C) have been investigated.

Application

2-Methylbutane may be used in the following studies:
  • As starting reagent for the synthesis of isoprene.
  • Preparation of C5 and C6 alkyl nitrates.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-59.8 °F - closed cup

Flash Point(C)

-51 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johnson AW.
Invitation to Organic Chemistry., 338-338 (1999)
Johnson AW.
Invitation to Organic Chemistry., 50-54 (1999)
Eagleson M.
Concise Encyclopedia Chemistry, 560-560 (1994)
Rotational Isomerism and Thermodynamic Functions of 2-Methylbutane and 2, 3-Dimethylbutane. Vapor Heat Capacity and Heat of Vaporization of 2-Methylbutane.
Scott DW, et al.
Journal of the American Chemical Society, 73(4), 1707-1712 (1951)
Formation of alkyl nitrates from the reaction of branched and cyclic alkyl peroxy radicals with NO.
Atkinson R, et al.
International Journal of Chemical Kinetics, 16(9), 1085-1101 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service