Skip to Content
Merck
All Photos(1)

Key Documents

Y0001317

Testosterone enantate for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Testosterone enanthate, (17β)-17-[(1-Oxoheptyl)oxy]androst-4-en-3-one, 17β-Hydroxy-4-androsten-3-one 17-enanthate, 4-Androsten-17β-ol-3-one 17-enanthate, NSC 17591, Testosterone 17β-heptanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H40O3
CAS Number:
Molecular Weight:
400.59
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

testosterone

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C26H40O3/c1-4-5-6-7-8-24(28)29-23-12-11-21-20-10-9-18-17-19(27)13-15-25(18,2)22(20)14-16-26(21,23)3/h17,20-23H,4-16H2,1-3H3/t20-,21-,22-,23-,25-,26-/m0/s1

InChI key

VOCBWIIFXDYGNZ-IXKNJLPQSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Testosterone enantate for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Carrasco et al.
Journal of hepatology, 1(6), 573-578 (1985-01-01)
We report a case of multiple hepatic adenomas developed in a 32-year-old man with renal allograft after long-term therapy with testosterone enanthate. Histological assessment showed a benign hepatocellular adenoma, which has not regressed over a 6-month follow-up since androgen withdrawal.
Gerhard Binder et al.
Clinical endocrinology, 82(1), 100-105 (2014-09-11)
The distinction between constitutional delay of growth and puberty (CDGP) and isolated hypogonadotropic hypogonadism (IHH) in males with delayed puberty is difficult but important for timely treatment. We assessed the accuracy of the GnRH agonist test (triptorelin 0·1 mg) in
Animesh N Sharma et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 306(9), R627-R635 (2014-02-28)
How sex steroids modulate glucocorticoid feedback on the hypothalamic-pituitary-corticotrope (HPC) unit is controversial in humans. We postulated that testosterone (T) in men and estradiol (E2) in women govern unstressed cortisol-mediated negative feedback on ACTH secretion. To test this hypothesis, 24

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service