Skip to Content
Merck
All Photos(1)

Key Documents

W308302

Sigma-Aldrich

Triethyl citrate

natural, ≥99%, FG

Synonym(s):

Ethyl citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOC2H5)(CH2COOC2H5)2
CAS Number:
Molecular Weight:
276.28
FEMA Number:
3083
Beilstein:
1801199
EC Number:
Council of Europe no.:
11762
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
09.512
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

vapor density

9.7 (vs air)

vapor pressure

1 mmHg ( 107 °C)

Assay

≥99%

refractive index

n20/D 1.442 (lit.)

bp

235 °C/150 mmHg (lit.)

mp

-55 °C

density

1.14 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

odorless

SMILES string

CCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC

InChI

1S/C12H20O7/c1-4-17-9(13)7-12(16,11(15)19-6-3)8-10(14)18-5-2/h16H,4-8H2,1-3H3

InChI key

DOOTYTYQINUNNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethyl citrate is an ester of citric acid, commonly used as an emulsifier and flavor preservative in the food industry. It is recognized as a safe direct food additive.

Application

Triethyl citrate is used as a food additive to stabilize foams, particularly egg whites.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

311.0 °F - closed cup

Flash Point(C)

155 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cast Natural Rubber Films Comprising Triethyl Citrate
Issarayungyuen P, et al.
Advanced Materials Research, 844, 494-497 (2014)
Safety assessment of citric acid, inorganic citrate salts, and alkyl citrate esters as used in cosmetics
Fiume MM, et al.
International Journal of Toxicology, 33(Supplement 2), 16S-46S (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service