Skip to Content
Merck
All Photos(1)

Key Documents

SMB01385

Sigma-Aldrich

SAICAR

≥95% (HPLC)

Synonym(s):

N-[[5-amino-1-(5-O-phosphono-β-D-ribofuranosyl)-1H-imidazol-4-yl]carbonyl]-L-aspartic acid, Succino-AICAR, Succinyl-AICAR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19N4O12P
CAS Number:
Molecular Weight:
454.28
UNSPSC Code:
12352202
NACRES:
NA.21

biological source

synthetic

Quality Level

Assay

≥95% (HPLC)

form

solid

application(s)

metabolomics

shipped in

2-8°C

storage temp.

2-8°C

SMILES string

NC1=C(N=CN1[C@H]2[C@H](O)[C@H](O)[C@@H](COP(O)(O)=O)O2)C(N[C@@H](CC(O)=O)C(O)=O)=O

General description

SAICAR is an intermediate in purine metabolism. SAICAR is converted from 5-amino-1-(5-phospho-D-ribosyl) imidazole-4-carboxylate (CAIR, Carboxy-AIR, carboxyaminoimidazole ribotide; CAS 6001-14-5) by phosphoribosylaminoimidazole-succinocarboxamide synthase (SAICAR synthase). SAICAR (a ribotide) can lose its phosphate group leading to the appearance of a riboside known as succinylaminoimidazolecarboxamide riboside (SAICAriboside or SAICAr) in cerebrospinal fluid, in urine, and, to a lesser extent, in plasma. When present in sufficiently high levels, SAICAR can act as an oncometabolite, where it promotes tumour growth & survival under glucose-limited conditions by stimulating pyruvate kinase isoform M2 (PKM2) and promoting aerobic glycolysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A mild form of adenylosuccinate lyase deficiency in absence of typical brain MRI features diagnosed by whole exome sequencing
Macchiaiolo Marina, et al.
Italian Journal of Pediatrics, 43, 65-65 (2017)
Extended diagnosis of purine and pyrimidine disorders from urine: LC MS/MS assay development and clinical validation
Monostori Peter, et al.
PLoS ONE, 14, e0212458-e0212458 (2019)
Ming Yan et al.
Biochemistry, 55(33), 4731-4736 (2016-08-03)
Human pyruvate kinase isoform M2 (PKM2) is a glycolytic enzyme isoform implicated in cancer. Malignant cancer cells have higher levels of dimeric PKM2, which is regarded as an inactive form of tetrameric pyruvate kinase. This perceived inactivity has fueled controversy
Kirstie E Keller et al.
Molecular cell, 53(5), 700-709 (2014-03-13)
Abnormal metabolism and sustained proliferation are hallmarks of cancer. Pyruvate kinase M2 (PKM2) is a metabolic enzyme that plays important roles in both processes. Recently, PKM2 was shown to have protein kinase activity phosphorylating histone H3 and promoting cancer cell
Roxane Marsac et al.
Genetics, 211(4), 1297-1313 (2019-02-01)
Purine homeostasis is ensured through a metabolic network widely conserved from prokaryotes to humans. Purines can either be synthesized de novo, reused, or produced by interconversion of extant metabolites using the so-called recycling pathway. Although thoroughly characterized in microorganisms, such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service