Skip to Content
Merck
All Photos(1)

Key Documents

PHR1116

Supelco

L-Menthol

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(−)-Menthol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, 5-Methyl-2-(1-methylethyl)cyclohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
Beilstein:
1902293
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 772
traceable to Ph. Eur. M0350000
traceable to USP 1381709

API family

menthol

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

InChI

1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m1/s1

InChI key

NOOLISFMXDJSKH-KXUCPTDWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

L-Menthol may be used as a pharmaceutical reference standard for the determination of the analyte in topical formulations by liquid and gas chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0294 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H G Grigoleit et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 12(8), 612-616 (2005-08-27)
The principal pharmacodynamic effect of peppermint oil relevant to the gastrointestinal tract is a dose-related antispasmodic effect on the smooth musculature due to the interference of menthol with the movement of calcium across the cell membrane. The choleretic and antifoaming
H F Stich et al.
Cancer letters, 15(3), 193-202 (1982-03-01)
Saliva of volunteers chewing betel quid, cured betel nut (Areca catechu), betel leaves (Piper betle), a mixture of quid ingredients (dried betel nut flakes, catechu, cardamon, lime, copra and menthol) and Indian tobacco was collected and examined for its genotoxic
Robert J Stein et al.
The Journal of urology, 172(3), 1175-1178 (2004-08-18)
Overactive bladder symptoms due to various etiologies have been successfully treated with capsaicin by desensitization of the temperature sensitive vanilloid receptor TRPV1. Recently another temperature sensitive receptor, TRPM8, activated by menthol and cool temperatures (8C to 28C) was described that
Tibor Rohács et al.
Nature neuroscience, 8(5), 626-634 (2005-04-27)
The subjective feeling of cold is mediated by the activation of TRPM8 channels in thermoreceptive neurons by cold or by cooling agents such as menthol. Here, we demonstrate a central role for phosphatidylinositol 4,5-bisphosphate (PI(4,5)P(2)) in the activation of recombinant
A Gelal et al.
Clinical pharmacology and therapeutics, 66(2), 128-135 (1999-08-25)
Menthol is widely used in a variety of commercial products and foods, but its clinical pharmacology is not well studied. To determine the disposition kinetics and to examine subjective and cardiovascular effects of menthol, we conducted a crossover placebo-controlled study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service